Skip to Content
Merck
All Photos(3)

Key Documents

P3886

Sigma-Aldrich

Poly-L-proline

suitable for ligand binding assays, Mol wt >30,000

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Poly-L-proline, mol wt >30,000

form

powder

Quality Level

mol wt

>30,000

technique(s)

ligand binding assay: suitable

color

white to light yellow

storage temp.

−20°C

SMILES string

OC(=O)C1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)

InChI key

ONIBWKKTOPOVIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Bysell et al.
Langmuir : the ACS journal of surfaces and colloids, 25(1), 522-528 (2008-12-09)
The relative importance of electrostatic and nonelectrostatic interactions in peptide-microgel systems was evaluated by micromanipulator-assisted light microscopy, confocal microscopy, and circular dichroism. For this purpose, the interaction of various homopolypeptides with lightly cross-linked polyelectrolyte gel particles ( approximately 70 microm
M Ashtari et al.
Journal of chromatography. A, 1265, 70-87 (2012-10-17)
Poly-proline chains and derivatives have been recently examined as the basis for new chiral stationary phases in high performance liquid chromatography. The selectivity of poly-proline has been measured for peptides with up to ten proline units. In this article, we
Biochemistry. Getting past polyproline pauses.
Allen R Buskirk et al.
Science (New York, N.Y.), 339(6115), 38-39 (2013-01-05)
Christopher J Woolstenhulme et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), E878-E887 (2013-02-23)
Although the ribosome is a very general catalyst, it cannot synthesize all protein sequences equally well. For example, ribosomes stall on the secretion monitor (SecM) leader peptide to regulate expression of a downstream gene. Using a genetic selection in Escherichia
Michael A Purdy
Journal of virology, 86(18), 10186-10193 (2012-07-20)
The hepatitis E virus (HEV) polyproline region (PPR) is an intrinsically unstructured region (IDR). This relaxed structure allows IDRs, which are implicated in the regulation of transcription and translation, to bind multiple ligands. Originally the nucleotide variability seen in the

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service