
Poly-L-proline

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Amino Acid
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing