Skip to Content
Merck
All Photos(4)

Key Documents

P2393

Sigma-Aldrich

Polyoxyethylene (10) tridecyl ether

mixture of C11 to C14 iso-alkyl ethers with C13 iso-alkyl predominating

Synonym(s):

C13E10

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

description

non-ionic

SMILES string

OCCCCCCCCCCCCC.OCCO

InChI key

QOXZVZPASZVADM-UHFFFAOYSA-N

General description

Polyoxyethylene (10) tridecyl ether (POE TDA) is a nonionic surfactant. [1]

Application

Polyoxyethylene (10) tridecyl ether has been used in a study to assess the role of polyribosomes and the cytoskeleton in protein accumulation of maize endosperm. [2] It has also been used in a study to investigate the application of commercial polyoxyethylene detergents used with the membrane protein phosphatidylinositol kinase.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein accumulation in the maize endosperm: role of polyribosomes and the cytoskeleton
Abe et al.
Plant Physiology and Biochemistry, 41, 7-7 (2003)
Application of hydrophilic-lipophilic balance (HLB) number to optimize a compatible non-ionic surfactant for dried aerial conidia of Beauveria bassiana
Jin et al.
Biological Control, 46, 8-8 (2008)
M I Aveldaño
Archives of biochemistry and biophysics, 324(2), 331-343 (1995-12-20)
The solubilization of rhodopsin and phospholipids from disks prepared from bovine retinal rods was studied using five different detergents. The relative amounts of rhodopsin and lipid extracted during membrane solubilization differed dramatically with the nature of the surfactant; the two
J Giudicelli et al.
Biochimica et biophysica acta, 831(1), 59-66 (1985-09-20)
Neutral alpha-D-glucosidase (alpha-D-glucoside glucohydrolase, EC 3.2.1.20) from horse kidney brush-border membranes was solubilized using Emulphogene BC 720 and purified by an affinity chromatography technique. The enzyme preparation (390-fold purified), which was free of other known microvillus hydrolases, exhibited one precipitate
Simultaneous demonstration of phagocytosis-connected oxygen consumption and corresponding NAD(P)H oxidase activity: direct evidence for NADPH as the predominant electron donor to oxygen in phagocytizing human neutrophils.
M Nakamura et al.
Biochemical and biophysical research communications, 98(3), 743-751 (1981-02-12)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service