Skip to Content
Merck
All Photos(5)

Key Documents

107700

Sigma-Aldrich

4-(Dimethylamino)pyridine

ReagentPlus®, ≥99%

Synonym(s):

N,N-Dimethylpyridin-4-amine, DMAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Beilstein:
110354
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

chips
crystalline powder
flakes

concentration

≤100%

pH

(10,23 at -0,1 g/l at 22  °C)

bp

162 °C

mp

108-110 °C (lit.)

solubility

H2O: 50 mg/mL

functional group

amine

storage temp.

room temp

SMILES string

CN(C)c1ccncc1

InChI

1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3

InChI key

VHYFNPMBLIVWCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Dimethylamino)pyridine (DMAP) is a highly versatile nucleophilic catalyst for acylation reactions and esterifications. It is also employed in various organic transformations like Baylis-Hillman reaction, Dakin-West reaction, protection of amines, C-acylations, silylations, applications in natural products chemistry, and many others.

Application

4-(Dimethylamino)pyridine can be used as a catalyst:
  • To synthesize 3,5-disubstituted 2,6-dicyanoaniline by reacting malononitrile, aldehydes, and β-nitroolefins.
  • For the acylation of alcohols with acid anhydrides under auxiliary base- and solvent-free conditions to synthesize corresponding esters.
  • In Baylis-Hillman reaction to form carbon-carbon bond by the coupling of an activated alkene with an aldehyde or ketone.
A highly efficient catalyst for acylation reactions

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

255.2 °F

Flash Point(C)

124 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Iida et al.
Organic letters, 8(23), 5215-5218 (2006-11-03)
[Structure: see text] A powerful Ti-crossed Claisen condensation between ketene silyl acetals (KSAs) and acid chlorides was successfully performed to give alpha-monoalkylated esters and thermodynamically unfavorable (less accessible) alpha,alpha-dialkylated beta-keto esters in good yield (46 examples; 41-98% yield). A closely
A M van Wijk et al.
Analytical and bioanalytical chemistry, 400(5), 1375-1385 (2011-03-30)
A generic LC-MS/MS method was developed for the analysis of potentially genotoxic alkyl halides. A broad selection of alkyl halides were derivatized using 4-dimethylaminopyridine in acetonitrile. The reaction conditions for derivatization, i.e., solvent, reaction time, temperature and concentration of alkyl
Hangxiang Wang et al.
Journal of the American Chemical Society, 133(31), 12220-12228 (2011-07-19)
Catalysts hold promise as tools for chemical protein modification. However, the application of catalysts or catalyst-mediated reactions to proteins has only recently begun to be addressed, mainly in in vitro systems. By radically improving the affinity-guided DMAP (4-dimethylaminopyridine) (AGD) catalysts
Chenghu Yan et al.
Biomacromolecules, 10(8), 2013-2018 (2009-09-03)
An effective method for grafting L-lactide (LA) from unmodified cellulose by ring-opening polymerization (ROP) in homogeneous mild conditions is presented. By using 4-dimethylaminopyridine (DMAP) as an organic catalyst, cellulose-graft-poly(L-lactide) (cellulose-g-PLLA) copolymers with a molar substitution (MS(PLLA)) of PLLA in a
Min Suk Shim et al.
Journal of controlled release : official journal of the Controlled Release Society, 133(3), 206-213 (2008-11-11)
To maximize therapeutic effects, targeted delivery of nucleic acids (e.g., DNA and RNA) in their appropriate intracellular targets is highly desirable. In this study, primary amines of a model polymeric nonviral carrier, polyethylenimine (PEI), at two molecular weights (0.8 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service