Skip to Content
Merck
All Photos(1)

Key Documents

388858

Sigma-Aldrich

Brij® C10

average Mn ~683

Synonym(s):

Polyethylene glycol hexadecyl ether, Polyoxyethylene (10) cetyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H33(OCH2CH2)nOH, n~10
CAS Number:
Beilstein:
6450627
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~683

mp

32-34 °C (lit.)

density

0.977 g/mL at 25 °C (lit.)

HLB

12

SMILES string

CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C56H114O21/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-58-19-21-60-23-25-62-27-29-64-31-33-66-35-37-68-39-41-70-43-45-72-47-49-74-51-53-76-55-56-77-54-52-75-50-48-73-46-44-71-42-40-69-38-36-67-34-32-65-30-28-63-26-24-61-22-20-59-18-16-57/h57H,2-56H2,1H3

InChI key

NLMKTBGFQGKQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Brij® C10 is a surfactant that can be used for applications such as: improving the wetting of PEDOT:PSS which can further be used in the fabrication of thin film transistors, formation of reverse micelles for the synthesis of high yielding nanostructures, in the preparation of platinum electrode for fuel cell applications

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrodeposited Mesoporous Pt for Direct Ethanol Fuel Cells Anodes
Doan N
International Journal of Electrochemical Science, 11(4), 7631-7643 (2016)
Strategy for preparation of transparent organic thin film transistors with PEDOT:PSS electrodes and a polymeric gate dielectric
Albrecht G, et al.
Materials Science in Semiconductor Processing, 40, 772-776 (2015)
Scalable synthesis of sub-100 nm hollow carbon nanospheres for energy storage applications
Zhao H, et al.
Nano Research, 11(4), 1822-1833 (2018)
Sreekhar Cheboyina et al.
International journal of pharmaceutics, 359(1-2), 167-173 (2008-05-23)
A novel freeze pelletization technique was evaluated for the preparation of wax-based sustained release matrix pellets. Pellets containing water-soluble drugs were successfully prepared using a variety of waxes. The drug release significantly depended on the wax type used and the
Xue Li et al.
Chemosphere, 78(6), 752-759 (2009-12-17)
Currently, soil contamination by polybrominated diphenyl ethers (PBDEs) at e-waste recycling sites in China is of significant environmental concern and requires the identification of appropriate remediation technologies. In this paper, photodestruction of a model PBDE congener (BDE-99) in selected UV/surfactant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service