Skip to Content
Merck
All Photos(4)

Documents

O3880

Sigma-Aldrich

Sodium oleate

≥95% (capillary GC)

Synonym(s):

cis-9-Octadecenoic acid sodium salt, Oleic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COONa
CAS Number:
Molecular Weight:
304.44
Beilstein:
4046357
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

Olea europaea

Quality Level

Assay

≥95% (capillary GC)

form

powder

mp

232-235 °C (lit.)

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].CCCCCCCC\C=C/CCCCCCCC([O-])=O

InChI

1S/C18H34O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;

InChI key

BCKXLBQYZLBQEK-KVVVOXFISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Oleic acid is used to investigate effects on apolipoprotein B synthesis, degradation and secretion in cultured cells.

Biochem/physiol Actions

Oleic acid increases hepatic secretion of apolipoprotein B100 in hepatocyte cell lines and in mice . Oleic acid inhibits apolipoprotein B100 secretion at higher physiologic doses .

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arno van der Weijden et al.
Nature communications, 11(1), 4800-4800 (2020-09-25)
Out-of-equilibrium molecular systems hold great promise as dynamic, reconfigurable matter that executes complex tasks autonomously. However, translating molecular scale dynamics into spatiotemporally controlled phenomena emerging at mesoscopic scale remains a challenge-especially if one aims at a design where the system
Zhengtao Liu et al.
American journal of translational research, 8(3), 1319-1338 (2016-05-18)
Epidemiological survey identified that the variant patatin-like phospholipase domain-containing protein 3 (PNPLA3) gene at I148M position exerts direct effect in promoting hepatocellular carcinoma (HCC) under extraneous oxidative stress by interaction with obesity. However, the mechanism is still unknown. HepG2 cells
Dylan T King et al.
Langmuir : the ACS journal of surfaces and colloids, 27(18), 11381-11393 (2011-08-16)
The prediction of surfactant phase behavior has applications in a wide range of areas. An accurate modeling of liquid phase behavior can aid our understanding of colloidal process or be used to design phases that respond in a defined way
Yoshikazu Kameshima et al.
International journal of pharmaceutics, 381(1), 34-39 (2009-08-04)
Mg-Al-ascorbic acid (ASA)-layered double hydroxides (ASA-LDHs) with Mg/Al=3 were synthesized by ion-exchange, coprecipitation and reconstruction methods. Composites with sodium oleate (SOA)/ASA-LDH were prepared by an ion-exchange method using various concentrations of SOA solutions. The (003) basal spacing of the ASA-LDHs
Branka Dejanović et al.
Chemistry and physics of lipids, 164(1), 83-88 (2010-11-16)
Dilute aqueous systems composed of sodium oleate micelles and sodium oleate/oleic acid vesicles were investigated as a function of pH by electron spin resonance spectroscopy with TEMPO-stearate TEMPO-stearamide as well as with a positively charged water soluble spin label, TEMPO-choline.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service