Skip to Content
Merck
All Photos(4)

Key Documents

C5038

Sigma-Aldrich

Sodium octanoate

≥99% (capillary GC)

Synonym(s):

Caprylic acid sodium salt, Octanoic acid sodium salt, Sodium caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COONa
CAS Number:
Molecular Weight:
166.19
Beilstein:
3597723
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Assay

≥99% (capillary GC)

form

powder

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].CCCCCCCC([O-])=O

InChI

1S/C8H16O2.Na/c1-2-3-4-5-6-7-8(9)10;/h2-7H2,1H3,(H,9,10);/q;+1/p-1

InChI key

BYKRNSHANADUFY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Hassan et al.
Journal of colloid and interface science, 362(1), 118-126 (2011-07-05)
Differential scanning calorimetry (DSC), circular dichroism (CD), difference spectroscopy (UV-vis), Raman spectroscopy, and small-angle X-ray scattering (SAXS) measurements have been performed in the present work to provide a quantitatively comprehensive physicochemical description of the complexation between bovine fibrinogen and the
Nina Dinjaski et al.
Biomaterials, 35(1), 14-24 (2013-10-08)
Biomaterial-associated infections represent a significant clinical problem, and treatment of these microbial infections is becoming troublesome due to the increasing number of antibiotic-resistant strains. Here, we report a naturally functionalized bacterial polyhydroxyalkanoate (PHACOS) with antibacterial properties. We demonstrate that PHACOS
Makoto Anraku et al.
Biochimica et biophysica acta, 1702(1), 9-17 (2004-09-29)
The capability of octanoate, N-acetyl-L-tryptophanate (N-AcTrp) and other ions of fatty acids and amino acids to stabilize human serum albumin (HSA) against thermal and oxidative stress was studied. Native-PAGE showed that octanoate, and more hydrophobic fatty acids anions, stabilizes the
Rosario Pignatello et al.
Materials (Basel, Switzerland), 12(6) (2019-03-17)
A biodegradable poly(3-R-hydroxyalkanoate) synthesized by Pseudomonas mediterranea was investigated as a biomaterial to obtain colloidal drug delivery systems. Using a nanoprecipitation method, nanoparticles with a mean size of 155 nm and a negative surface charge were formed. They can be
Andrea S Pereyra et al.
Journal of lipid research, 62, 100069-100069 (2021-03-25)
Long-chain fatty acid oxidation is frequently impaired in primary and systemic metabolic diseases affecting the heart; thus, therapeutically increasing reliance on normally minor energetic substrates, such as ketones and medium-chain fatty acids, could benefit cardiac health. However, the molecular fundamentals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service