N1538
Nobiletin
≥97%
Synonym(s):
2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-1-benzopyran-4-one, 3′,4′,5,6,7,8-Hexamethoxyflavone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H22O8
CAS Number:
Molecular Weight:
402.39
Beilstein:
360887
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥97%
form
powder
storage temp.
−20°C
SMILES string
COc1ccc(cc1OC)C2=CC(=O)c3c(OC)c(OC)c(OC)c(OC)c3O2
InChI
1S/C21H22O8/c1-23-13-8-7-11(9-15(13)24-2)14-10-12(22)16-17(25-3)19(26-4)21(28-6)20(27-5)18(16)29-14/h7-10H,1-6H3
InChI key
MRIAQLRQZPPODS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Nobiletin as a novel agent to enhance porcine in vitro embryo development and quality: This study explores the potential of nobiletin to improve embryo development and quality in porcine models, providing insights into its applications in reproductive biotechnology and animal science (Cajas YN, Martinez-Serrano CA, 2024 Apr 22) (source).
- Nobiletin alleviates cisplatin-induced ototoxicity: Research indicates that nobiletin can mitigate the ototoxic effects of cisplatin chemotherapy by activating autophagy and inhibiting ferroptosis, suggesting a new therapeutic avenue for protecting auditory health during cancer treatment (Song W, Lu Z, 2024 Apr 3) (source).
- Nobiletin targets SREBP1/ACLY to induce autophagy-dependent cell death of gastric cancer cells: This study reveals that nobiletin promotes autophagy-dependent cell death in gastric cancer cells by targeting the SREBP1/ACLY axis within the PI3K/Akt/mTOR signaling pathway, proposing a novel anticancer strategy (Chen M, Sun Q, 2024 Jun) (source).
- Recent advances in the therapeutic potential of nobiletin against respiratory diseases: A comprehensive review on the therapeutic applications of nobiletin in treating respiratory diseases, emphasizing its anti-inflammatory and antioxidant properties as mechanisms for intervention (Qin Y, Li J, 2024 Jun) (source).
Biochem/physiol Actions
Nobiletin suppresses the expression of matrix metalloproteinases 1, 3 and 9, which participate in the break down of the extracellular matrix during tumor metastasis. Nobiletin is also involved in the increased expression of tissue inhibitor of matrix metalloproteinase 1 further preventing extracellular matrix degradation during tumor invasion. This bioflavonoid is believed to achieve both of these functions by disrupting the signal transduction pathway of Ras-Raf-MEK.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Salma Malik et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 67(7-8), 427-433 (2015-05-24)
Cisplatin is an effective anti-cancer drug which causes remarkable toxicity to kidney by generating reactive oxygen species and by stimulating inflammatory and apoptotic pathway. Citrus flavonoid, like nobiletin has been reported to possess anti-oxidant, anti-inflammatory and anti-apoptotic properties. Hence, the
Satoru Horigome et al.
Bioscience, biotechnology, and biochemistry, 78(5), 851-860 (2014-07-19)
The rhizome of Kaempferia parviflora has been used in traditional Thai medicine. In this study, we identified and compared specific compounds from the hexane extract of K. parviflora with those from other Zingiberaceous plants by using gas chromatography-mass spectrometry. We
Yuan Hao et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 37(1), 306-320 (2015-09-01)
Nobiletin, a citrus flavonoid isolated from tangerines, alters ion transport functions in intestinal epithelia, and has antagonistic effects on eosinophilic airway inflammation of asthmatic rats. The present study examined the effects of nobiletin on basal short-circuit current (I(SC)) in a
Feng-Quan Tan et al.
BMC plant biology, 15, 89-89 (2015-04-08)
Polyploidy has often been considered to confer plants a better adaptation to environmental stresses. Tetraploid citrus rootstocks are expected to have stronger stress tolerance than diploid. Plenty of doubled diploid citrus plants were exploited from diploid species for citrus rootstock
Su-Chen Ho et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 71, 176-182 (2014-06-24)
Inhibiting microglial activation-mediated neuroinflammation has become a convincing target for the development of functional foods to treat neurodegenerative diseases. Tangerine peel (Citri reticulatae pericarpium) has potent anti-inflammatory capacity; however, its anti-neuroinflammatory capacity and the corresponding active compounds remain unclear. To
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service