Skip to Content
Merck
All Photos(3)

Key Documents

H5254

Sigma-Aldrich

Hesperidin

≥80% (HPLC)

Synonym(s):

3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside, Hesperetin 7-rhamnoglucoside, Hesperitin-7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34O15
CAS Number:
Molecular Weight:
610.56
Beilstein:
75140
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

Citrus sinensis (L.) Osbeck

Assay

≥80% (HPLC)

form

powder

color

yellow to faint beige

mp

250-255 °C (dec.) (lit.)

solubility

soluble, yellow to brown

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

InChI

1S/C28H34O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-7,10,17,19,21-30,32-37H,8-9H2,1-2H3/t10-,17-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

QUQPHWDTPGMPEX-QJBIFVCTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Studies of the behavior of hesperidin and hesperetin (the aglycone) with DMPC liposomes find the aglycone penetrates more deeply and binds more strongly to the "membrane." This may explain the better bioavailability of bioflavonoid aglycones in general.

Biochem/physiol Actions

Flavonoid phytochemical extracted from citrus species. It is the rhamnoglucoside (rutinoside) of hesperetin.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yomna S Mo'men et al.
Journal of biochemical and molecular toxicology, e22305-e22305 (2019-02-20)
Hesperidin is a flavanone glycoside that is found in the Citrus species and showed antioxidant, hepatoprotective as well as anticancer activity. This study investigated the effect of hesperidin on the PI3K/Akt pathway as a possible mechanism for its protective effect
Yong Yang et al.
European journal of medicinal chemistry, 58, 390-395 (2012-11-17)
We have identified a previously unknown function of the natural compound, hesperetin. Here, we demonstrate that this small molecular agent is able to inhibit the transforming growth factor-β (TGF-β) signaling pathway. Single-molecule force measurements and single-molecule fluorescence imaging show that
Julia M Assini et al.
Current opinion in lipidology, 24(1), 34-40 (2012-12-21)
Citrus flavonoids are polyphenolic compounds with powerful biological properties. This review aims to summarize recent advances towards understanding the ability of citrus flavonoids to regulate lipid metabolism and other metabolic parameters relevant to the metabolic syndrome, type 2 diabetes and
Leandro C Souza et al.
Progress in neuro-psychopharmacology & biological psychiatry, 40, 103-109 (2012-09-22)
The present study investigated a possible antidepressant-like activity of hesperidin using two predictive tests for antidepressant effect in mice: the forced swimming test (FST) and the tail suspension test (TST). Results demonstrated that hesperidin (0.1, 0.3 and 1 mg/kg, intraperitoneal
Binit Kumar et al.
Vascular pharmacology, 57(5-6), 201-207 (2012-09-13)
The purpose of the study was to evaluate vasculoprotective effects of Hesperetin (Hsp) in Streptozotocin induced diabetic rats. The study was carried out for a period of 24weeks and evaluated for angiogenic parameters (VEGF and PKC-β), retinal vascular leakage by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service