Skip to Content
Merck
All Photos(1)

Key Documents

W431300

Sigma-Aldrich

Hesperetin

≥95%

Synonym(s):

3′,5,7-Trihydroxy-4′-methoxyflavanone, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
FEMA Number:
4313
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.097
NACRES:
NA.21

biological source

synthetic

Assay

≥95%

form

powder (with a light tan cast)

mp

227-232 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

COc1ccc(cc1O)C2CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H14O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-6,14,17-19H,7H2,1H3

InChI key

AIONOLUJZLIMTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuan Zeng et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1136, 121846-121846 (2019-12-11)
Naringin has been documented to possess multiple pharmacological activities. Reported pharmacokinetic studies revealed that oral bioavailability of naringin was low, in contrast to its significant pharmacological effects. The in vivo distribution of naringin and derived metabolites might partly explain this
Elena Valeria Fuior et al.
Pharmaceutics, 11(8) (2019-08-07)
Citrus flavonoids have well-documented protective effects on cardiovascular system, but the poor water solubility and reduced bioavailability restrict their therapeutic use. We aimed to overcome these limitations and encapsulated naringenin and hesperetin into lipid nanoemulsions (LNs), targeted to vascular cell
Ramesh Elango et al.
Journal of Asian natural products research, 20(6), 559-569 (2017-05-26)
We studied the chemoprevention property of hesperetin on H522 cells using MTT, an apoptosis assay, an analysis of cell cycle progression, and the mitochondrial membrane potential, and apoptotic marker gene expression was determined using quantitative PCR. Hesperetin enhanced apoptotic cell
Chuye Ji et al.
International journal of biological macromolecules, 168, 775-783 (2020-11-24)
The interaction between biomacromolecules and ligands has attracted great interest because of their biological properties. Calf thymus DNA (ctDNA) can interact with bioactive compounds to form complexes. Here, ctDNA-ligand complexes were studied using fluorescence, absorption, and infrared spectroscopy, circular dichroism
Ebubekir Izol et al.
Chemistry & biodiversity, 18(1), e2000560-e2000560 (2020-11-15)
Allium species are widely consumed as food all over the world. The phenolic profile of ethanol extracts of aerial parts and roots of 12 Allium species, collected from five different Eastern Anatolia regions, were studied using LC-MS/MS. In vitro antioxidant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service