Skip to Content
Merck
All Photos(2)

Key Documents

MTOX1097P24

Sigma-Aldrich

MRP7 Knockout Caco-2 Cells

human cervix, Epithelial

Synonym(s):

MRP7 Knockout C2BBe1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Product Name

MRP7 Knockout Caco-2 Cells, one assay ready, 24 well plate

biological source

human colon

form

liquid

morphology

Epthelial

technique(s)

drug transporter assay: suitable
permeability assay: suitable

application(s)

ADME/TOX

Looking for similar products? Visit Product Comparison Guide

General description

The C2BBe1 cells, a subclone of Caco-2 cells, correspond to ATCC Cat. No. CRL-2102. The MRP7 knockout C2BBe1 cells are adenocarcinoma, epithelial cells from a human caucasian male (aged 72 years) with functional knockout of the MRP7 efflux transporter.

The three week production lead time begins on the Monday following a purchase, in the third week the cells are shipped overnight for receipt on Tuesday or Wednesday. As a biologic product that′s shipped at room temperature the cells must be processed immediately upon receipt.

Features and Benefits

The Caco-2 subclone, C2BBe1 cells, are ideal for transporter analysis as they express multiple transporters, are human derived and grow in a homogenous monolayer that forms tight juntions which is necessary for efflux ratio analysis. Other benefits include:

- A functional knockout of the MRP7 gene eliminates the reliance on chemical inhibitors to determine if a compound is an MRP7 substrate
- The 24 well Transwell format enables the MRP7 knockout cells to be included in standard drug transporter protocols
- Human assay with no interference from animal inhibitors
- Overcome the limitations of RNAi and knockdown cell lines that arise from remaining transporter functionality

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Artursson
Journal of pharmaceutical sciences, 79(6), 476-482 (1990-06-01)
A human intestinal cell line, Caco-2, was used as a model to study the passive diffusion of drugs across intestinal epithelium. The cells formed continuous monolayers when grown on permeable filters of polycarbonate. After 10 days in culture, the monolayers
Mark I Kaldas et al.
The Journal of pharmacy and pharmacology, 55(3), 307-312 (2003-05-02)
Resveratrol is a dietary constituent suggested to have protective effects against cancer as well as cardiovascular disease. The purpose of the study was to learn whether this agent could be absorbed in man and enter the systemic circulation. This was
S Yee
Pharmaceutical research, 14(6), 763-766 (1997-06-01)
To evaluate and optimize the use of Caco-2 cell monolayers to predict the in vivo absorption of a broad range of compounds in man. Caco-2 cells are derived from human adenocarcinoma colon cells and spontaneously differentiate when grown on porous
X Wu et al.
Pharmaceutical research, 17(2), 209-215 (2000-04-06)
The purpose of this study was to elucidate the mechanisms by which an HMG-CoA reductase inhibitor, atorvastatin (an organic acid with a pKa of 4.46), was transported in the secretory and absorptive directions across Caco-2 cell monolayers. Caco-2 cells were
V Pade et al.
Journal of pharmaceutical sciences, 87(12), 1604-1607 (1999-04-03)
The objective of this investigation was to establish a relationship between drug permeability and solubility in vitro and the extent of drug absorption in humans. We selected drugs with varying permeabilities and solubilities with the aim of establishing a relationship

Articles

Application note on Drug transport assays in a 96-well system using Millicell-96 System from Millipore.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service