Skip to Content
Merck
All Photos(1)

Key Documents

09042001

Sigma-Aldrich

CACO-2

human cervix, Epithelial

Synonym(s):

Caco-II Cells, Caco2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CACO-2, Caucasian colon adenocarcinoma

biological source

human colon

Quality Level

form

liquid

growth mode

(Adherent)

karyotype

(Hypertetraploid, modal no. 96)

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human caucasian colon adenocarcinoma, intestinal permeability characteristics tested

Cell Line Description

Isolated from a primary colonic tumour in a 72-year-old Caucasian male using the explant culture technique. Forms moderately well differentiated adenocarcinomas consistent with colonic primary grade II, in nude mice. This CACO-2 catalogue number has undergone testing for intestinal permeability: Dome, microvilli and tight junction formation and transport functionality tested and confirmed present at ECACC.

Application

CACO-2 has been used in the cytotoxicity studies with high pressure-assisted extraction (HPE) fraction of nettle leaf extracts.
CACO-2 may be used as a cell model to study:
  • in vitro permeability assay with various biopharmaceuticals
  • in iron uptake and absorption studies
  • in the cytotoxicity assay with surfactants and transporter inhibitors
  • in liposome formulated drug uptake and cytotoxicity studies

Biochem/physiol Actions

Caco-2 (Cancer coli-2) is derived from colorectal adenocarcinoma and grows as an adherent monolayer of epithelial cells. Caco-2 cell line is a widely used intestinal epithelium model and has unique spontaneous differentiation and confluence property. It actively transports vitamins, hormones, amino acids and sugars. Caco-2 expresses various enzymes, ionic and non-ionic transporters similar to normal human epithelia. Caco-2 cell line models grow on filter support. They have simplicity and reproducibility and provide reliable information. Since Caco-2 mimics the intestinal epithelium, it is the preferred cell model for cytotoxic studies, food and drug transport studies.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 11
D13S317: 11,13,14
D16S539: 12,13
D5S818: 12,13
D7S820: 11,12
THO1: 6
TPOX: 9,11
vWA: 16,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum FBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm² using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. NB: During routine subculture the cells should always be subcultured before they achieve confluence. Cells may show the appearance of circular vacuoles in the cytoplasm. These may increase in frequency as the culture density increases to confluence. To reduce their frequency, media change confluent cultures after 2-3 days if not subcultured. Cells can clump if not separated into a single cell suspension when split.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
NaviCyte Scientific holds the exclusive commercial distribution rights to the CACO-2 cell line deposited by the Memorial Sloan-Kettering Cancer Center. Note: All uses of Catalogue Numbers 86010202 and 09042001, other than for research by a non-commercial or academic entity, require a license and use authorization from NaviCyte Scientific under its exclusive arrangement with Memorial Sloan-Kettering Cancer Center. For information on the licensing terms, please contact NaviCyte Scientific via [email protected] or (+1) 973-868-6100.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison study of oral iron preparations using a human intestinal model
Zariwala MG, et al.
Scientia Pharmaceutica, 81, 1123-1140 (2013)
Caco-2 Cell Line
Verhoeckx K, et al.
The Impact of Food Bioactives on Health, 103-111 (2015)
Araceli Valverde et al.
PloS one, 10(6), e0131363-e0131363 (2015-06-25)
Despite the demonstrated benefits of anti-EGFR/VEGF targeted therapies in metastatic colorectal cancer (mCRC), many patients initially respond, but then show evidence of disease progression. New therapeutic strategies are needed to make the action of available drugs more efficient. Our study
Biopharmaceutical classification of desloratadine-not all drugs are classified the easy way
Berginc K, et al.
Acta pharmaceutica (Zagreb, Croatia), 70, 131-144 (2020)
M Lautenschläger et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 22(1), 36-44 (2015-02-01)
Extracts of saffron (Crocus sativus L.) have traditionally been used against depressions. Recent preclinical and clinical investigations have rationalized this traditional use. Trans-crocetin, a saffron metabolite originating from the crocin apocarotenoids, has been shown to exert strong NMDA receptor affinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service