Skip to Content
Merck
All Photos(2)

Key Documents

MTOX1006P24

Sigma-Aldrich

BCRP/MRP2 Double Knockout Caco-2 Cells

human cervix, Epithelial

Synonym(s):

BCRP/MRP2 Double Knockout C2BBe1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

BCRP/MRP2 Double Knockout Caco-2 Cells, one assay ready, 24 well plate

biological source

human colon

form

solid

morphology

Epthelial

technique(s)

drug transporter assay: suitable
permeability assay: suitable

application(s)

ADME/TOX

Looking for similar products? Visit Product Comparison Guide

General description

The C2BBe1 cells, a subclone of Caco-2 cells, correspond to ATCC CRL-2102. The BCRP/MRP2 double knockout C2BBe1 cells are adenocarcinoma, epithelial cells from a human caucasian male (aged 72 years) with functional knockout of the BCRP and MRP2 efflux transporters.

The three week production lead time begins on the Monday following a purchase, in the third week the plates are shipped on Tuesday for receipt on Wednesday or Thursday. As a biologic product that′s shipped at room temperature the cells must be processed immediately upon receipt.

Features and Benefits

The Caco-2 subclone, C2BBe1 cells, are ideal for transporter analysis as they express multiple transporters, are human derived and grow in a homogenous monolayer that forms tight juntions which is necessary for efflux ratio analysis. Other benefits include:

  • Functional knockout of the BCRP and MRP2 genes eliminates the reliance on chemical inhibitors to determine if a compound is a BCRP or MRP2 substrate
  • The 24 well Transwell format enables the BCRP and MRP2 knockout cells to be included in standard drug transporter protocols
  • Human assay with no interference from animal inhibitors
  • Overcome the limitations of RNAi and knockdown cell lines that arise from remaining transporter functionality

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Artursson
Journal of pharmaceutical sciences, 79(6), 476-482 (1990-06-01)
A human intestinal cell line, Caco-2, was used as a model to study the passive diffusion of drugs across intestinal epithelium. The cells formed continuous monolayers when grown on permeable filters of polycarbonate. After 10 days in culture, the monolayers
V Pade et al.
Journal of pharmaceutical sciences, 87(12), 1604-1607 (1999-04-03)
The objective of this investigation was to establish a relationship between drug permeability and solubility in vitro and the extent of drug absorption in humans. We selected drugs with varying permeabilities and solubilities with the aim of establishing a relationship
X Wu et al.
Pharmaceutical research, 17(2), 209-215 (2000-04-06)
The purpose of this study was to elucidate the mechanisms by which an HMG-CoA reductase inhibitor, atorvastatin (an organic acid with a pKa of 4.46), was transported in the secretory and absorptive directions across Caco-2 cell monolayers. Caco-2 cells were
S Yee
Pharmaceutical research, 14(6), 763-766 (1997-06-01)
To evaluate and optimize the use of Caco-2 cell monolayers to predict the in vivo absorption of a broad range of compounds in man. Caco-2 cells are derived from human adenocarcinoma colon cells and spontaneously differentiate when grown on porous
M J Briske-Anderson et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 214(3), 248-257 (1997-03-01)
The Caco-2 cell line is used by many investigators as a model of the intestinal epithelium to study nutrient uptake and transport. Our goal was to create an awareness of inherent variabilities in the Caco-2 cell line which may influence

Articles

Application note on Drug transport assays in a 96-well system using Millicell-96 System from Millipore.

Utilize these Caco-2 cell based assay tools for screening small molecule drug compounds prior to clinical studies and submission to regulatory agencies.

Utilize these Caco-2 cell based assay tools for screening small molecule drug compounds prior to clinical studies and submission to regulatory agencies.

Utilize these Caco-2 cell based assay tools for screening small molecule drug compounds prior to clinical studies and submission to regulatory agencies.

See All

Related Content

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service