Skip to Content
Merck
All Photos(1)

Key Documents

MTOX1004

Sigma-Aldrich

MDR1/BCRP Double Knockout Caco-2 Cells

Human male colorectal tissue, adenocarcinoma

Synonym(s):

C2BBe1 Cells MDR1/BCRP (-/-/-/-,-/-/-/-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

Product Name

MDR1/BCRP Double Knockout Caco-2 Cells, one vial

biological source

human male colorectal tissue (Source disease: adenocarcinoma)

form

liquid

technique(s)

drug transporter assay: suitable
permeability assay: suitable

OMIM accession no.

application(s)

ADME/TOX

storage temp.

−196°C

Gene Information

General description

The C2BBe1 cells, a subclone of Caco-2 cells, correspond to ATCC CRL-2102. The MDR1 and BCRP knockout C2BBe1 cells are adenocarcinoma, epithelial cells from a human caucasian male (aged 72 years) with functional knockout of the ABCB1 and ABCG2 (MDR1/BCRP) efflux transporters.

Features and Benefits

The Caco-2 subclone C2BBe1 cells are ideal for transporter analysis as they express multiple transporters, are human derived, and grow in a homogenous monolayer that forms tight junctions necessary for efflux ratio analysis. Other benefits include:
  • A functional double knockout of the MDR1 gene and BCRP gene eliminates the reliance on chemical inhibitors to determine if a compound is an MRP2 substrate
  • The vial format enables the MDR1 and BCRP knockout cells to be included in standard drug transporter protocols
  • Human assay with no interference from animal inhibitors
  • Overcome the limitations of RNAi and knockdown cell lines that arise from remaining transporter functionality

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • MTOX1004MDR1/BCRP Double Knockout Caco-2 Cells, one vial 1 vialSDS

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Artursson
Journal of pharmaceutical sciences, 79(6), 476-482 (1990-06-01)
A human intestinal cell line, Caco-2, was used as a model to study the passive diffusion of drugs across intestinal epithelium. The cells formed continuous monolayers when grown on permeable filters of polycarbonate. After 10 days in culture, the monolayers
S Yee
Pharmaceutical research, 14(6), 763-766 (1997-06-01)
To evaluate and optimize the use of Caco-2 cell monolayers to predict the in vivo absorption of a broad range of compounds in man. Caco-2 cells are derived from human adenocarcinoma colon cells and spontaneously differentiate when grown on porous
X Wu et al.
Pharmaceutical research, 17(2), 209-215 (2000-04-06)
The purpose of this study was to elucidate the mechanisms by which an HMG-CoA reductase inhibitor, atorvastatin (an organic acid with a pKa of 4.46), was transported in the secretory and absorptive directions across Caco-2 cell monolayers. Caco-2 cells were
V Pade et al.
Journal of pharmaceutical sciences, 87(12), 1604-1607 (1999-04-03)
The objective of this investigation was to establish a relationship between drug permeability and solubility in vitro and the extent of drug absorption in humans. We selected drugs with varying permeabilities and solubilities with the aim of establishing a relationship
Mark I Kaldas et al.
The Journal of pharmacy and pharmacology, 55(3), 307-312 (2003-05-02)
Resveratrol is a dietary constituent suggested to have protective effects against cancer as well as cardiovascular disease. The purpose of the study was to learn whether this agent could be absorbed in man and enter the systemic circulation. This was

Articles

We presents an article on The Role of Intestinal Efflux Transporters In Drug Absorption.

We presents an article on The Role of Intestinal Efflux Transporters In Drug Absorption.

We presents an article on The Role of Intestinal Efflux Transporters In Drug Absorption.

We presents an article on The Role of Intestinal Efflux Transporters In Drug Absorption.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service