Skip to Content
Merck
All Photos(1)

Key Documents

M6768

Sigma-Aldrich

Methoxypolyethylene glycol 350

average mol wt 350

Synonym(s):

Polyethylene glycol, Methoxypolyethylene glycol, Poly(ethylene glycol) methyl ether, mono-Methyl polyethylene glycol 350

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nH
CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

mol wt

average mol wt 350

application(s)

microbiology

SMILES string

O(CCO)C

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

General description

Methoxypolyethylene glycol 350 is a polymer similar in structure and nomenclature to polyethylene glycols.

Application

Methoxypolyethylene glycol 350 has been used in a study to assess the synthesis of a new class of thermosensitive micellar cyclotriphosphazenes. It has also been used in a study to investigate synthesis of a new amphiphilic poly(organophosphazene) by stepwise nucleophilic substitution.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thermosensitive and biocompatible cyclotriphosphazene micelles
U, Toti.
J. Controlled Release, 119, 7-7 (2007)
S J Hermansky et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 35(10-11), 1031-1039 (1998-02-17)
The methoxy polyethylene glycols (MPEGs), also referred to as polyethylene glycol methyl ethers, are high molecular weight polymers similar in structure and nomenclature to the polyethylene glycols. Because of the potential for repeated cutaneous exposure of humans to MPEG-350 and
Synthesis, characterization, and tumor selectivity of a polyphosphazene-platinum(II) conjugate
Rita Song et al.
J. Controlled Release, 105, 9-9 (2005)
Pengxiang Zhao et al.
Chemical communications (Cambridge, England), 49(31), 3218-3220 (2013-03-14)
"Click" chemistry now offers access to a great variety of triazoles, and the first example of a strategy to stabilize gold nanoparticles (AuNPs) with a new 1,2,3-triazole-mPEG ligand is developed here together with preliminary examples of possible applications.
Lina Du et al.
Anti-cancer drugs, 24(2), 172-180 (2012-09-20)
A functionalized poly(amidoamine) (PAMAM) nanocarrier was designed and prepared to deliver anticancer drugs. The nanocarrier is a copolymer with a core-shell structure with 3.0 G PAMAM as the core and sequentially conjugated poly(2-(N,N-diethylamino)ethyl methacrylate) (pDEA) and methoxy-poly(ethylene glycol) 2000 (mPEG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service