Skip to Content
Merck
All Photos(1)

Key Documents

202487

Sigma-Aldrich

Poly(ethylene glycol) methyl ether

average MN 550, methoxy, hydroxyl

Synonym(s):

Polyethylene glycol, Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol) methyl ether, average Mn 550

vapor density

>1 (vs air)

vapor pressure

0.05 mmHg ( 20 °C)

form

semisolid

mol wt

average Mn 550

refractive index

n20/D 1.455

viscosity

7.5 cSt(210 °F)(lit.)

transition temp

Tm 20 °C

density

1.089 g/mL at 25 °C

Ω-end

hydroxyl

α-end

methoxy

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) methyl ether can be used as a pore-forming agent to prepare polysulfone membranes with enhanced hydrophilicity.

Poly(ethylene glycol) methyl ether-grafted polyamidoamine (PAMAM) dendrimers can be used as drug carrier systems for anticancer drugs.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pengxiang Zhao et al.
Chemical communications (Cambridge, England), 49(31), 3218-3220 (2013-03-14)
"Click" chemistry now offers access to a great variety of triazoles, and the first example of a strategy to stabilize gold nanoparticles (AuNPs) with a new 1,2,3-triazole-mPEG ligand is developed here together with preliminary examples of possible applications.
Yiyi Yu et al.
Journal of pharmaceutical sciences, 102(3), 1054-1062 (2013-01-03)
To promote the application of methoxy poly(ethylene glycol)-cholesterol (mPEG-Chol), mPEG-Chol was used to prepare core-shell micelles encapsulating poorly water-soluble docetaxel (DTX-PM) by modified cosolvent evaporation method. Approaches to enhance DTX entrapment efficiency (EE) and minimize particle size were investigated in
Hyo Won Seo et al.
Biomaterials, 34(11), 2748-2757 (2013-01-25)
The effectiveness of systemically administered anticancer treatments is limited by difficulties in achieving therapeutic doses within tumors, a problem that is complicated by dose-limiting side effects to normal tissue. To increase the efficacy and reduce the toxicity of systemically administered
Mulu Z Tesfay et al.
Journal of virology, 87(7), 3752-3759 (2013-01-18)
We are developing oncolytic vesicular stomatitis viruses (VSVs) for systemic treatment of multiple myeloma, an incurable malignancy of antibody-secreting plasma cells that are specifically localized in the bone marrow. One of the presumed advantages for using VSV as an oncolytic
Yanjie Bao et al.
Pharmaceutical research, 30(2), 342-351 (2012-09-18)
To determine the influence of physicochemical properties of lipid nanoparticles (LNPs) carrying siRNA on their gene silencing in vivo. Mechanistic understanding of how the architecture of the nanoparticle can alter gene expression has also been studied. The effect of 3-N-[(ω-methoxypoly(ethylene

Articles

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service