Skip to Content
Merck
All Photos(1)

Key Documents

07964

Sigma-Aldrich

Methoxypolyethylene glycol amine

750

Synonym(s):

mPEG-NH2, O-(2-Aminoethyl)-O′-methylpolyethylene glycol, Aminopolyethylene glycol monomethyl ether, Methoxypolyoxyethylene amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2(OCH2CH2)nOCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.22

Quality Level

form

solid

color

white to yellow

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methoxypolyethylene glycol amine can:
  • Form dendritic micelles to enhance the solubility of hydrophobic compounds.
  • Conjugate with an anticancer drug methotrexate (MTX) to form MTX-PEG with potential application as drug carriers.
  • Couple with polyethylenimine (PEI) to form graft copolymers which can be used as nonviral gene vectors.

Other Notes

Oligomer used to prepare PEG-ylated conjugates having the advantage of reduced immunogenicity and increased serum half life

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of polyethylene glycol on gene delivery of polyethylenimine.
Sung SJ, et al.
Biological & Pharmaceutical Bulletin, 26(4), 492-500 (2003)
Polyethylene glycol-polyamidoamine dendritic micelle as solubility enhancer and the effect of the length of polyethylene glycol arms on the solubility of pyrene in water.
Yang H, et al.
Journal of Colloid and Interface Science, 273(1), 148-154 (2004)
Polyethylene Glycol Conjugates of Methotrexate Varying in Their Molecular Weight from MW 750 to MW 40000: Synthesis, Characterization, and Structure- Activity Relationships in Vitro and in Vivo.
Riebeseel K, et al.
Bioconjugate Chemistry, 13(4), 773-785 (2002)
T.P. Kogan
Synthetic Communications, 22, 2417-2417 (1992)
M Yokoyama et al.
Bioconjugate chemistry, 3(4), 295-301 (1992-07-01)
Adriamycin, a hydrophobic anticancer drug, was conjugated with poly(ethylene oxide)-poly(aspartic acid) block copolymers composed of various lengths of each block copolymer segment ranging from 1000 to 12,000 in molecular weight and from 10 to 80 units, respectively. Conjugation was achieved

Articles

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service