Skip to Content
Merck
All Photos(3)

Key Documents

M006

Sigma-Aldrich

Proglumide sodium salt

solid

Synonym(s):

4-Benzoylamino-5-dipropylamino-5-oxopentanoic acid sodium-potassium salt, N-Benzoyl-N′,N′-dipropyl-DL-isoglutamine sodium-potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25N2NaO4
CAS Number:
Molecular Weight:
356.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: soluble

SMILES string

[Na+].CCCN(CCC)C(=O)C(CCC([O-])=O)NC(=O)c1ccccc1

InChI

1S/C18H26N2O4.Na/c1-3-12-20(13-4-2)18(24)15(10-11-16(21)22)19-17(23)14-8-6-5-7-9-14;/h5-9,15H,3-4,10-13H2,1-2H3,(H,19,23)(H,21,22);/q;+1/p-1

InChI key

UFMWGCINVOIJSO-UHFFFAOYSA-M

Gene Information

Application

Proglumide sodium salt has been used to study the effects of proglumide on macronutrient selection in European sea bass Dicentrarchus labrax, L.

Biochem/physiol Actions

Proglumide sodium salt is a non-peptide cholecystokinin receptor antagonist that has greater selectivity for the CCKA subtype. It selectively blocks central nervous system effects. Proglumide sodium salt is orally active.
CCK antagonist; selectively blocks CNS effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The cholecystokinin antagonist, proglumide, increases food intake in the rat
G.Shillabeer and J. Davidson
Regulatory Peptides (1984)
E S Corp et al.
Physiology & behavior, 61(6), 823-827 (1997-06-01)
Cholecystokinin (CCK) receptors are classified as two subtypes, designated CCK(A) and CCK(B), and both subtypes are found in brain and peripheral tissues of rats. CCK-8 has been shown to act peripherally to reduce meal size, and this satiating action can
J V Menani et al.
The American journal of physiology, 275(5 Pt 2), R1431-R1437 (1998-10-29)
The present study investigated the effects of bilateral injections of the nonselective CCK receptor antagonist proglumide or CCK-8 into the lateral parabrachial nuclei (LPBN) on the ingestion of 0.3 M NaCl and water induced by intracerebroventricular injection of ANG II
Role of cholecystokinin and its antagonist proglumide on macronutrient selection in European sea bass Dicentrarchus labrax, L.
V. C. Rubio
Physiology & Behavior (2008)
James M Hill et al.
Science translational medicine, 6(265), 265ra169-265ra169 (2014-12-05)
Herpesviruses are highly prevalent and maintain lifelong latent reservoirs, thus posing challenges to the control of herpetic disease despite the availability of antiviral pharmaceuticals that target viral DNA replication. The initiation of herpes simplex virus infection and reactivation from latency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service