Skip to Content
Merck
All Photos(4)

Key Documents

L9908

Sigma-Aldrich

LY-294,002 hydrochloride

from microbial (Staphylococcus roseus), ≥98% (HPLC), solid, PI3K inhibitor

Synonym(s):

2-(4-Morpholinyl)-8-phenyl-1(4H)-benzopyran-4-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO3 · HCl
CAS Number:
Molecular Weight:
343.80
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

LY-294,002 hydrochloride, solid, ≥98% (HPLC)

biological source

microbial (Staphylococcus roseus)

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >5 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

O=C1C=C(Oc2c1cccc2-c3ccccc3)N4CCOCC4

InChI

1S/C19H17NO3/c21-17-13-18(20-9-11-22-12-10-20)23-19-15(7-4-8-16(17)19)14-5-2-1-3-6-14/h1-8,13H,9-12H2

InChI key

CZQHHVNHHHRRDU-UHFFFAOYSA-N

General description

LY-294,002 hydrochloride is a derivative of the flavonoid, quercetin and elicits higher inhibition on the enzyme phosphoinositide 3-kinase (PI3K).[1] LY-294,002 inhibits nuclear factor kappa B signaling in macrophages.[2] It elevates expression of autophagosomal protein LC3 and promotes apoptosis in gastric[3] and nasopharyngeal cancer cells.[4] LY-294,002 modulates action potential repolarization and improves myocyte contractility.[5]

Application

LY-294,002 hydrochloride has been used in the inhibition of:
  • phosphoinositide 3-kinase (PI3K) in human lung cancer cell line[6]
  • autophagy in mesenchymal stem cells (MSCs).[7]
  • serine/threonine-specific protein kinase (AKT) signaling in fibroblasts[8] and cardiac cells.[9]

Biochem/physiol Actions

Specific cell permeable phosphatidylinositol 3-kinase inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide and Kinase Phosphatase Biology research. Discover more featured Cyclic Nucleotide and Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the G Proteins (Heterotrimeric) and Phosphoinositide Kinases pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carboxyl-Terminal Modulator Protein Ameliorates Pathological Cardiac Hypertrophy by Suppressing the Protein Kinase B Signaling Pathway
Liu X, et al.
Journal of the American Heart Association, 7(13), e008654-e008654 (2018)
miR-126 suppresses epithelial-to-mesenchymal transition and metastasis by targeting PI3K/AKT/Snail signaling of lung cancer cells
Jia Z, et al.
Oncology Letters, 15(5), 7369-7375 (2018)
Structural determinants of phosphoinositide 3-kinase inhibition by wortmannin, LY294002, quercetin, myricetin, and staurosporine
Walker EH, et al.
Molecular Cell, 6(4), 909-919 (2000)
C J Vlahos et al.
Journal of immunology (Baltimore, Md. : 1950), 154(5), 2413-2422 (1995-03-01)
Neutrophils contain a multicomponent NADPH oxidase system that is involved in the production of microbicidal oxidants. Stimulation of human neutrophils with the peptide FMLP activates this respiratory burst enzyme to produce superoxide and also has been shown to result in
G J Brunn et al.
The EMBO journal, 15(19), 5256-5267 (1996-10-01)
The immunosuppressant, rapamycin, inhibits cell growth by interfering with the function of a novel kinase, termed mammalian target of rapamycin (mTOR). The putative catalytic domain of mTOR is similar to those of mammalian and yeast phosphatidylinositol (PI) 3-kinases. This study

Articles

We presents an article on Autophagy in Cancer Promotes Therapeutic Resistance

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

See All

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service