Skip to Content
Merck
All Photos(1)

Key Documents

SML0428

Sigma-Aldrich

GSK690693

≥98% (HPLC)

Synonym(s):

4-(2-(4-Amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-{[(3S)-3-piperidinylmethyl]oxy}-1H-imidazo[4,5-c]pyridin-4-yl)-2-methyl-3-butyn-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O3
CAS Number:
Molecular Weight:
425.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CCn1c(nc2c(ncc(OC[C@H]3CCCNC3)c12)C#CC(C)(C)O)-c4nonc4N

InChI

1S/C21H27N7O3/c1-4-28-18-15(30-12-13-6-5-9-23-10-13)11-24-14(7-8-21(2,3)29)16(18)25-20(28)17-19(22)27-31-26-17/h11,13,23,29H,4-6,9-10,12H2,1-3H3,(H2,22,27)/t13-/m0/s1

InChI key

KGPGFQWBCSZGEL-ZDUSSCGKSA-N

Application

GSK690693 has been used in western blotting and immunoprecipitations.[1]

Biochem/physiol Actions

4-(2-(4-Amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-{[(3S)-3-piperidinylmethyl]oxy}-1H-imidazo[4,5-c]pyridin-4-yl)-2-methyl-3-butyn-2-ol/GSK690693[2] is a protein kinase B/AKT inhibitor. GSK690693 can prevent growth and apoptosis in acute lymphoblastic leukemia cell lines.[3]
GSK690693 is a potent pan-AKT kinase inhibitor.
GSK690693 is an ATP competitive, potent pan-AKT kinase inhibitor with IC50 values of 2, 13, and 9 nM against AKT1, 2, and 3, respectively.

Features and Benefits

This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The IKK/NF-kappaB signaling pathway requires Morgana to drive breast cancer metastasis
Fusella F, et al.
Nature Communications, 8(1), 1636-1636 (2017)
Identification of 4-(2-(4-Amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-
Heerding D A, et al.
Journal of medicinal chemistry, 51(18), 5663-5679 (2008)
Michael K Kiessling et al.
PloS one, 11(1), e0147682-e0147682 (2016-01-29)
High-risk neuroblastoma remains lethal in about 50% of patients despite multimodal treatment. Recent attempts to identify molecular targets for specific therapies have shown that Neuroblastoma RAS (NRAS) is significantly mutated in a small number of patients. However, few inhibitors for
Masayuki Yamaji et al.
Cancer medicine, 6(11), 2646-2659 (2017-09-30)
Malignant pleural mesothelioma (MPM), an asbestos-related occupational disease, is an aggressive and incurable tumor of the thoracic cavity. Despite recent advances in MPM treatment, overall survival of patients with MPM is very low. Recent studies have implicated that PI3K/Akt signaling
AKT inhibitor, GSK690693, induces growth inhibition and apoptosis in acute lymphoblastic leukemia cell lines
Levy D S, et al.
Blood, 113(8), 1723-1729 (2009)

Articles

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service