Skip to Content
Merck
All Photos(1)

Key Documents

L2778

Sigma-Aldrich

Latex beads, amine-modified polystyrene, fluorescent red

aqueous suspension, 1.0 μm mean particle size

Synonym(s):

Fluorescent latex beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.56
Pricing and availability is not currently available.

form

aqueous suspension

composition

Solids, 2.5%

technique(s)

cell based assay: suitable

mean particle size

1.0 μm

fluorescence

λex ~575 nm; λem ~610 nm

application(s)

cell analysis

Looking for similar products? Visit Product Comparison Guide

Application

Latex beads, amine-modified polystyrene, fluorescent red has been used:
  • to study the effect of Ganoderma lucidum polysaccharides (GLP) on microglia phagocytic behavior[1]
  • to study cell phagocytosis in human leukemia cell lines[2]
  • in macrophage phagocytosis assay[3]
  • to induce detectable engulfment by microglia in Ganoderma lucidum[1]

Biochem/physiol Actions

Amine-modified polystyrene latex beads have been used to develop an electrochemical nitrite nanosensor as well as to validate a pharyngeal aspiration technique for exposing the mouse lung to respirable particles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa D Brown et al.
Parasites & vectors, 9, 237-237 (2016-04-28)
Vector-borne pathogens must overcome arthropod infection and escape barriers (e.g. midgut and salivary glands) during the extrinsic incubation period (EIP) before subsequent transmission to another host. This particular timespan is undetermined for the etiological agent of flea-borne spotted fever (Rickettsia
Ya Xu et al.
Oncology reports, 37(5), 2711-2719 (2017-04-26)
Microvesicle biogenesis is a highly regulated process. Aberrant release of microvesicles from cancer cells have been associated with their invasiveness and prognosis. However, the mechanism of aberrant release remains poorly understood. Herein, we found that hepatocellular carcinoma cells shed more
Qing Cai et al.
Journal of neuroinflammation, 14(1), 63-63 (2017-03-28)
Ganoderma lucidum (GL) has been widely used in Asian countries for hundreds of years to promote health and longevity. The pharmacological functions of which had been classified, including the activation of innate immune responses, suppression of tumour and modulation of
Huan Yang et al.
Frontiers in immunology, 9, 1987-1987 (2018-09-21)
Urinary tract infections (UTIs) caused by uropathogenic Escherichia coli (UPEC) induce cystitis, pyelonephritis, and can cause kidney scarring and failure if inflammation is not under control. The detailed effects of cytotoxic necrotizing factor 1 (CNF1), the key UPEC toxin, on
Anirban Choudhury et al.
Chest, 131(3), 809-815 (2007-03-16)
Platelet microparticles (PMPs), are procoagulant membrane vesicles that are derived from activated platelets, the levels of which are elevated in patients with hypertension, coronary artery disease (CAD), diabetes, and stroke, all of which are conditions that lead to (and are

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service