Skip to Content
Merck

LB1

Latex beads, polystyrene

0.1 μm mean particle size

Synonym(s):

Latex Beads, Latex Microspheres, laboratory latex beads, latex sphers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous suspension

composition

Solids, 10%

packaging

pack of 1 ea

mean particle size

0.1 μm

General description

Polystyrene microparticles are anionic-stabilizedcolloidal particles. These microparticles are synthesized by the polymerizationof styrene under conditions that produce coalescent bead formation. Polystyrenelatex beads have a wide range of applications including phagocytosis experiments,antibody-mediated agglutination diagnostics, electron microscopy, and cellcounter calibration.

Application

Polystyrene latex beads have been used:
  • inthe flow cytometric assay of endothelial microparticles (EMPs)
  • in thedetection of capsular polysaccharide (CPS) antibody/antigen latex agglutinationtest
  • to evaluate the phagocytic activity in hemocytes of shrimps


Still not finding the right product?

Explore all of our products under Latex beads, polystyrene


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Evaluation of Cellular Absorption and Metabolism of ??Carotene Loaded in Nanocarriers after In Vitro Digestion
Ling C, et al.
Journal of Agricultural and Food Chemistry, 9383-9394, 69-69 (2021)
Francesca Capetti et al.
Journal of separation science, 43(9-10), 1879-1889 (2020-02-20)
The frankincense resins, secreted from Boswellia species, are an uncommon example of a natural raw material where every class of terpenoids is present in similar proportions. Diterpenoids (serratol, incensole, and incensole acetate) are used to discriminate samples from different species
The effect of MWCNTs on molar mass in in situ polymerization of styrene and methyl methacrylate
Annala M, et al.
European Polymer Journal, 48(9), 1516-1524 (2012)



Global Trade Item Number

SKUGTIN
LB1-2ML04061838208415
LB1-1ML04061838108203
LB1-15ML04061838208408