Skip to Content
Merck
All Photos(1)

Key Documents

L9654

Sigma-Aldrich

Latex beads, amine-modified polystyrene, fluorescent orange

aqueous suspension, 1.0 μm mean particle size

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.56

form

aqueous suspension

composition

Solids, 2.4-2.6%

technique(s)

cell based assay: suitable

mean particle size

1.0 μm

fluorescence

λex ~520 nm; λem ~540 nm

application(s)

cell analysis

Looking for similar products? Visit Product Comparison Guide

Application

Amine-modified polystyrene latex beads (fluorescent orange) have been used to develop an electrochemical nitrite nanosensor as well as to validate a pharyngeal aspiration technique for exposing the mouse lung to respirable particles.
They have also been used:
  • to test nanofiber filtration efficiency
  • in the preparation of bilayer tissue-simulating phantoms, which are artificial materials used in biomedical optical instrumentation
  • to copolymerize collagen coated stiff polyacrylamide gels to seed cancer cells for traction force microscopy

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recycled PET nanofibers for water filtration applications
Zander N, et al.
Materials, 9(4), 247-247 (2016)
Development of thin skin mimicking bilayer solid tissue phantoms for optical spectroscopic studies
Nivetha KB and Sujatha N
Biomedical Optics Express, 8(7), 3198-3212 (2017)
Development of thin skin mimicking bilayer solid tissue phantoms for optical spectroscopic studies.
K Bala Nivetha et al.
Biomedical optics express, 8(7), 3198-3212 (2017-07-19)
MMP proteolytic activity regulates cancer invasiveness by modulating integrins
Das A, et al.
Scientific reports, 7(1), 14219-14219 (2017)
Ronald Lulijwa et al.
Journal of fish biology, 96(3), 731-746 (2020-01-30)
Polyinosinic:polycytidylic acid [poly (I:C)] was administered in vivo to Chinook salmon (Oncorhynchus tshawytscha) post-smolts to determine the immune responses on haematological and cellular functional parameters, including spleen (SP), head kidney (HK) and red blood cell (RBC) cytokine expression, as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service