Skip to Content
Merck
All Photos(3)

Key Documents

I9516

Sigma-Aldrich

2-Propanol

2-Propanol

≥99.5%, for molecular biology, suitable for RNA extraction, DNA extraction, BioReagent

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.52
grade:
for molecular biology
technique(s):
DNA extraction: suitable
RNA extraction: suitable
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

2-Propanol, for molecular biology, BioReagent, ≥99.5%

grade

for molecular biology

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

product line

BioReagent

Assay

≥99.5%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

DNA extraction: suitable
RNA extraction: suitable

impurities

≤0.2% water
water

color

colorless

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble (completely)

density

0.785 g/mL at 25 °C (lit.)

suitability

suitable for precipitation of nucleic acids

storage temp.

room temp

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropanol is a clear, colorless, polar organic solvent commonly used in chemistry and molecular biology laboratories. It will dissolve a wide range of chemicals, and evaporates quickly. This product (2-propanol) is designated as molecular biology grade and is suitable for the precipitation of nucleic acids.

Protocols for the use of isopropanol:

  • DNA precipitation from mammalian tissue, ethidium bromide extraction from DNA samples, and RNA isolation from cells and tissues.[1]
  • Protein crystallization.[2][3][4]
  • Isolate retinoids from biological matrices for HPLC and HPLC-MS analysis.[5][6]

Application

2-Propanol has been used:
  • in DNA and RNA extraction[7][8][9][10]
  • for determination of hydroxyproline levels(The Merck Index, 12th ed)
  • for checking cell viability by MTT assay[11]
  • in microarray hybridization[12]
Suitable for use in DNA precipitation using standard protocols. Also suitable as a solvent for making solutions for molecular biology applications.

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert B Nicewonger et al.
Bioorganic & medicinal chemistry letters, 12(14), 1799-1802 (2002-06-28)
In this paper, we describe the synthesis of N-(6-cyano-1,3-dimethyl-2,4-dioxo-5-substituted-1,3-dihydropyridino[2,3-d] pyrimidin-7-yl)imides 1. We will show the synthesis of 1 using both conventional heating and microwave techniques. In addition, the imide was attached to polystyrene and this immobilized imide was equally as
S Knapp et al.
Proteins, 23(4), 595-597 (1995-12-01)
The thermostable amylopullulanase from Pyrococcus woesei was crystallized. Crystals, suitable for a crystallographic analysis up to a size of 0.6 mm in their longest dimension, have been obtained by the vapor diffusion method in a solution containing polyethyleneglycol 4000 (PEG
Christudas Morais et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 25(5), 1462-1474 (2009-12-29)
Renal cell carcinoma (RCC) is a highly metastatic and lethal disease with few efficacious treatments. Many studies have shown that the ubiquitous transcription factor nuclear factor kappa B (NF-kappaB) plays a key role in the development and progression of many
M J Romão et al.
European journal of biochemistry, 215(3), 729-732 (1993-08-01)
The Desulfovibrio gigas aldehyde oxidoreductase contains molybdenum bound to a pterin cofactor and [2Fe-2S] centers. The enzyme was characterized by SDS/PAGE, gel-filtration and analytical ultracentrifugation experiments. It was crystallized at 4 degrees C, pH 7.2, using isopropanol and MgCl2 as
Peter K Davidsen et al.
Journal of applied physiology (Bethesda, Md. : 1985), 110(2), 309-317 (2010-10-30)
MicroRNAs (miRNA), small noncoding RNA molecules, may regulate protein synthesis, while resistance exercise training (RT) is an efficient strategy for stimulating muscle protein synthesis in vivo. However, RT increases muscle mass, with a very wide range of effectiveness in humans.

Articles

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

Protocols

Procedure and protocol for Anti Ago-RNA Immunoprecipitation from mammalian cells using the RIP kit

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

See All

Related Content

Isopropanol is a polar organic solvent that is widely used in chemistry, biochemistry and molecular biology

Reviews

Active Filters

  1. 4 Ratings-only reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service