Skip to Content
Merck
2-Propanol BioRenewable, ReagentPlus®, ≥99.5%

2-Propanol

2-Propanol BioRenewable, ReagentPlus®, ≥99.5%
Synonym(s):
sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol, 2-Propanol
Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-20 of 40
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2