Skip to Content
Merck
All Photos(1)

Key Documents

I8283

Sigma-Aldrich

ICI 63,197

≥98% (HPLC)

Synonym(s):

2-Amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5(4H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5O
CAS Number:
Molecular Weight:
207.23
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white

mp

161.8-163 °C

solubility

DMSO: 13 mg/mL

storage temp.

2-8°C

SMILES string

CCCN1C(=O)C(C)=Cn2nc(N)nc12

InChI

1S/C9H13N5O/c1-3-4-13-7(15)6(2)5-14-9(13)11-8(10)12-14/h5H,3-4H2,1-2H3,(H2,10,12)

InChI key

UQDVRVNMIJAGRK-UHFFFAOYSA-N

Biochem/physiol Actions

Phosphodiesterase IV (PDE 4) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Wachtel
Psychopharmacology, 77(4), 309-316 (1982-01-01)
The significance of a characteristic symptomatology (hypothermia, hypoactivity, forepaw shaking, grooming, head twitches) as a potential in vivo correlate of enhanced availability of brain adenosine cyclic 3',5'-monophosphate (cAMP) was examined in rats following systemic administration of various doses of dibutyryladenosine
S P Canfield et al.
International journal of tissue reactions, 5(3), 241-243 (1983-01-01)
The effect of 5 hydroxytryptamine (5-HT) on gastric acid secretion by the rat isolated stomach has been studied. 5-HT (10 microM) significantly inhibited secretory responses to pentagastrin, histamine and isoprenaline but not those due to bethanechol, dibutyryl cyclic AMP or
H O Collier et al.
Federation proceedings, 40(5), 1513-1518 (1981-04-01)
A quasi-morphine withdrawal syndrome (QMWS) is a pattern of behavior closely resembling the true withdrawal syndrome in the opiate-dependent animal, which can be elicited acutely by a nonopiate drug in an opiate-naive animal. The main criteria proposed for the QMWS
J Abrahamsen et al.
Naunyn-Schmiedeberg's archives of pharmacology, 343(2), 161-165 (1991-02-01)
The aim of the present investigation was to examine whether or not presynaptic facilitatory beta-adrenoceptors are detectable on the postganglionic nerves in the rabbit isolated ear artery. Strips of rabbit central ear artery were incubated with 3H-noradrenaline (10(-7) mol/l; 30
C B Archer et al.
The British journal of dermatology, 109(5), 559-564 (1983-11-01)
Peripheral blood lymphocyte cyclic AMP responses to isoprenaline, prostaglandin E2 and histamine were examined in patients with atopic eczema, in the presence and absence of a potent phosphodiesterase inhibitor (PDEI). Basal cyclic AMP levels were not significantly different in atopic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service