Skip to Content
Merck
All Photos(2)

Key Documents

36763

Supelco

Clofentezine

PESTANAL®, analytical standard

Synonym(s):

3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine, Clofentezine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8Cl2N4
CAS Number:
Molecular Weight:
303.15
Beilstein:
7137128
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

182-185 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccccc1-c2nnc(nn2)-c3ccccc3Cl

InChI

1S/C14H8Cl2N4/c15-11-7-3-1-5-9(11)13-17-19-14(20-18-13)10-6-2-4-8-12(10)16/h1-8H

InChI key

UXADOQPNKNTIHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Riga et al.
Scientific reports, 7(1), 9202-9202 (2017-08-25)
The mechanisms underlying insecticide and acaricide resistance in insects and mites are often complex, including additive effects of target-site insensitivity, increased metabolism and transport. The extent to which target-site resistance mutations contribute to the resistance phenotype is, however, not well
Dingxu Li et al.
Experimental & applied acarology, 38(4), 255-273 (2006-04-14)
A leaf disc bioassay was employed in the laboratory to assess the population responses of Tetranychus viennensis Zacher to clofentezine at three concentrations (LC(10), LC(25) and LC(50)) in three successive generations. Life-table parameters were calculated in each acaricide exposure regime
K A Barnes et al.
Rapid communications in mass spectrometry : RCM, 9(14), 1441-1445 (1995-01-01)
A high performance liquid chromatographic/atmospheric pressure chemical ionization-mass spectrometric (HPLC/APCI-MS) method has been developed for the determination of the pesticides diflubenzuron (1-(4-chlorophenyl)-3-(2,6-difluorobenzoyl)urea) and clofentezine (3,6-bis(2-chlorophenyl)-1,2,4,5-tetrazine) in plums, strawberries and blackcurrant-based fruit drinks. Samples were homogenized with acetone, extracted into dichloromethane
Tomasz Tuzimski
Journal of separation science, 33(13), 1954-1958 (2010-05-28)
The aim of this study is to report a new procedure for extraction, cleanup and determination of clofentezine in herb extracts by ultrasound-assisted solvent extraction, SPE and multidimensional planar chromatography with diode array detector (MDPC-DAD) and/or HPLC-DAD. The application of
Xiuyan Li et al.
Journal of hazardous materials, 304, 474-480 (2015-12-01)
A simple and highly sensitive aptamer-based colorimetric sensor was developed for selective detection of Microcystin-LR (MC-LR). The aptamer (ABA) was employed as recognition element which could bind MC-LR with high-affinity, while gold nanoparticles (AuNPs) worked as sensing materials whose plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service