Skip to Content
Merck
All Photos(5)

Documents

D216305

Sigma-Aldrich

2,2′-Bipyridyl

greener alternative

ReagentPlus®, ≥99%

Synonym(s):

2,2′-Bipyridine, 2,2′-Dipyridine, 2,2′-Dipyridyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2
CAS Number:
Molecular Weight:
156.18
Beilstein:
113089
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder or crystals
powder or granules

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

273 °C (lit.)

mp

70-73 °C (lit.)

solubility

ethanol: 100 mg/mL, clear, colorless to faintly yellow

greener alternative category

storage temp.

room temp

SMILES string

c1ccc(nc1)-c2ccccn2

InChI

1S/C10H8N2/c1-3-7-11-9(5-1)10-6-2-4-8-12-10/h1-8H

InChI key

ROFVEXUMMXZLPA-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2,2′-Bipyridyl also known as 2,2′-bipyridine, is a symmetrical bipyridine commonly used as a neutral ligand for chelating with metal ions. It is a bidentate ligand which has been employed transition metal catalysis, aluminum initiated polymerization, and various other inorganic syntheses. This compound adheres to the principles of greener chemistry, ensuring catalytic efficiency. Click here for more information

Application

2,2′-Bipyridyl has been used in:
  • the preparation of fluorescent dyes like products 754730, 544981, 804215.
  • the preparation ofcopper(II)-bipyridine-naringenin complex.
  • the preparation of 2,2′-bipyridyl hydrobromide.
  • as a ligand in the greener oxidation of alcohols under aerobic conditions
  • as a complex with CuBR2 in the green synthesis of propargylamines
  • the synthesis of [Ru(bipy)3]Cl2, a common metal complex with usefulphotochemistry often used in photoredox catalysis as a sensitizer

Biochem/physiol Actions

Metalloprotease inhibitor, high-affinity chelator of iron; may inhibit Fe2+ containing enzymes at 10−8 M.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

267.8 °F - closed cup

Flash Point(C)

131 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromination of 2,2'-bipyridile.
Zdravkov AB and Khimich NN.
Russ. J. Org. Chem., 42(8), 1200-1202 (2006)
Macromolecules, 27, 645-645 (1994)
Tetrahedron Letters, 34, 6219-6219 (1993)
Júlio César Conceição Filho et al.
PloS one, 9(9), e107058-e107058 (2014-09-06)
Cancer is the second leading cause of death worldwide and there is epidemiological evidence that demonstrates this tendency is emerging. Naringenin (NGEN) is a trihydroxyflavanone that shows various biological effects such as antioxidant, anticancer, anti-inflammatory, and antiviral activities. It belongs
Coordination Chemistry Reviews, 93, 205-205 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service