Skip to Content
Merck
All Photos(4)

Key Documents

H3128

Sigma-Aldrich

Calconcarboxylic acid

suitable for use as an indicator for the titration of calcium

Synonym(s):

2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acid, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2-carboxylic acid, Patton and Reeder’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14N2O7S
CAS Number:
Molecular Weight:
438.41
Beilstein:
727492
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
application(s):
diagnostic assay manufacturing
hematology
histology

form

powder

mp

300 °C

solubility

1 M NaOH: 10 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)c1cc2ccccc2c(N=Nc3c(O)cc(c4ccccc34)S(O)(=O)=O)c1O

InChI

1S/C21H14N2O7S/c24-16-10-17(31(28,29)30)13-7-3-4-8-14(13)18(16)22-23-19-12-6-2-1-5-11(12)9-15(20(19)25)21(26)27/h1-10,24-25H,(H,26,27)(H,28,29,30)/b23-22-

InChI key

MVQBFZXBLLMXGS-FCQUAONHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calconcarboxylic acid is a useful indicator for calcium titrations with EDTA in the presence of magnesium. It is also required for the staining of proteins in gels.
Calconcarboxylic acid is also known as Patton-Reeder indicator and is used in the determination of calcium at pH 12. Calconcarboxylic acid is a chelating agent and is used in the preparation of chelating sorbent along with chemically modified silica LiChroprep-NH2, for trace metal analysis. Calconcarboxylic acid coupled with 3-aminopropyltriethoxysilane (APS-CCA), a fluorescent probe, facilitates the medical detection of Ca2+ ions in the blood with minimal blood backgrounds using hydrophobic hexadecyltrimethoxysilane (HDS) as substrate in fluorimetric microarray method.

Application

Calconcarboxylic acid (CCA) has been used in the preparation of poly-CCA film for leukemia diagnosis by immobilization, hybridization of DNA and detection by methylene blue and differential pulse voltammetry.

Quality

Contains an inert diluent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation Methods In Proteomics (2005)
A high-throughput fluorimetric microarray with enhanced fluorescence and suppressed ?coffee-ring? effects for the detection of calcium ions in blood
Ding Y, et al.
Scientific Reports, 6, 38602-38602 (2016)
Javier Villela-Castrejón et al.
Journal of food science, 82(7), 1726-1734 (2017-06-10)
Corn lime cooking generates a large amount of wastewater known as nejayote that is composed of suspended solids and solubilized phytochemicals. Spray drying can be an alternative to recover bioactive molecules, such as ferulic acid, from nejayote. Besides the yield
An electrochemical biosensor for detection of PML/RARA fusion gene using capture probe covalently immobilized onto poly-calcon carboxylic acid modified glassy carbon electrode
Wei N, et al.
Talanta, 78(4-5), 1227-1234 (2009)
Patton, J. and Reeder, W.
Analytical Chemistry, 28, 1026-1026 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service