Skip to Content
Merck
All Photos(4)

Key Documents

G9003

Sigma-Aldrich

D-Glutamine

≥98% (HPLC)

Synonym(s):

D-2-Aminoglutaramic acid, D-Glutamic acid 5-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O3
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723796
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Product Name

D-Glutamine, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

application(s)

cell analysis

SMILES string

N[C@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m1/s1

InChI key

ZDXPYRJPNDTMRX-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutamine is a non-essential and the most abundant free amino acid present in the human body. It is an important component of proteins and is one of the 20 proteinogenic amino acids. It forms 5-6% of bound amino acids.

Application

D-Glutamine has been used to study its role in conferring protection against acetaldehyde-induced disruption of barrier function in Caco-2 cell monolayer.

Biochem/physiol Actions

Glutamine forms the central metabolite in amino acid transamination via a-ketoglutarate and glutamic acid. This amino acid is metabolized by different enzymes, such as glutaminase, present in liver, and glutamine synthetase, present in skeletal muscle. It is produced in the cytoplasm from other amino acids, predominantly from branched-chain amino acids and glutamate. It plays an essential role in ammonia metabolism and detoxification. Its skeletal muscle levels are significantly reduced post trauma, operation and inflammatory states. It servers as a prognostic marker in fatal sepsis during which its skeletal muscle levels are decreased by 90%.

Other Notes

Unnatural isomer of glutamine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Anesi et al.
Materials (Basel, Switzerland), 13(10) (2020-05-21)
(1) Background: a cell evaluation focused to verify the self-regenerative antioxidant activity is performed on cerium doped bioactive glasses. (2) Methods: the glasses based on 45S5 Bioglass®, are doped with 1.2 mol%, 3.6 mol% and 5.3 mol% of CeO2 and
Indra Saptiama et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(18), 4843-4855 (2019-01-18)
The effective utilization of various biomolecules for creating a series of mesoporous boehmite (γ-AlOOH) and gamma-alumina (γ-Al2 O3 ) nanosheets with unique hierarchical multilayered structures is demonstrated. The nature and concentration of the biomolecules strongly influence the degree of the
Erich Roth
The Journal of nutrition, 138(10), 2025S-2031S (2008-09-23)
Glutamine is the most abundant free amino acid of the human body. Besides its role as a constituent of proteins and its importance in amino acid transamination, glutamine has regulatory capacity in immune and cell modulation. Glutamine deprivation reduces proliferation
Mingbang Wang et al.
mSystems, 4(1) (2019-02-01)
Changes in the gut microenvironment may influence the pathogenesis of autism spectrum disorders (ASD). Here, we investigated the composition of the gut microbiota and metabolites in children with ASD. Ninety-two children with ASD and 42 age-matched children exhibiting typical development
G Thorsén et al.
Journal of chromatography. B, Biomedical sciences and applications, 745(2), 389-397 (2000-10-24)
A method is presented for the chiral analysis of amino acids in biological fluids using micellar electrokinetic chromatography (MEKC) and laser-induced fluorescence (LIF). The amino acids are derivatized with the chiral reagent (+/-)-1-(9-anthryl)-2-propyl chloroformate (APOC) and separated using a mixed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service