
D-Glutamine

Synonym(s):
D-2-Aminoglutaramic acid, D-Glutamic acid 5-amide
Empirical Formula (Hill Notation):
C5H10N2O3
CAS Number:
Molecular Weight:
146.14
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1