Merck
  • Cell Proliferation to Evaluate Preliminarily the Presence of Enduring Self-Regenerative Antioxidant Activity in Cerium Doped Bioactive Glasses.
Cell Proliferation to Evaluate Preliminarily the Presence of Enduring Self-Regenerative Antioxidant Activity in Cerium Doped Bioactive Glasses.
Materials (Basel, Switzerland) (2020-05-21)
Alexandre Anesi, Gianluca Malavasi, Luigi Chiarini, Roberta Salvatori, Gigliola Lusvardi
ABSTRACT
(1) Background: a cell evaluation focused to verify the self-regenerative antioxidant activity is performed on cerium doped bioactive glasses. (2) Methods: the glasses based on 45S5 Bioglass®, are doped with 1.2 mol%, 3.6 mol% and 5.3 mol% of CeO2 and possess a polyhedral shape (~500 µm2). Glasses with this composition inhibit oxidative stress by mimicking catalase enzyme (CAT) and superoxide dismutase (SOD) activities; moreover, our previous cytocompatibility tests (neutral red (NR), 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) and Bromo-2-deoxyUridine (BrdU)) reveal that the presence of cerium promotes the absorption and vitality of the cells. The same cytocompatibility tests were performed and repeated, in two different periods (named first and second use), separated from each other by four months. (3) Results: in the first and second use, NR tests indicate that the presence of cerium promotes once again cell uptake and viability, especially after 72 h. A decrease in cell proliferation it is observed after MTT and BrdU tests only in the second use. These findings are supported by statistically significant results (4) Conclusions: these glasses show enhanced proliferation, both in the short and in the long term, and for the first time such large dimensions are studied for this kind of study. A future prospective is the implantation of these bioactive glasses as bone substitute in animal models.
Sigma-Aldrich
D-Glutamine, ≥98% (HPLC)