Skip to Content
Merck
All Photos(1)

Key Documents

G6416

Sigma-Aldrich

GW5074

powder

Synonym(s):

3-(3,5-Dibromo-4-hydroxybenzyliden)-5-iodo-1,3-dihydroindol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8Br2INO2
CAS Number:
Molecular Weight:
520.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow to orange-brown

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

SMILES string

Oc1c(Br)cc(cc1Br)\C=C2/C(=O)Nc3ccc(I)cc23

InChI

1S/C15H8Br2INO2/c16-11-4-7(5-12(17)14(11)20)3-10-9-6-8(18)1-2-13(9)19-15(10)21/h1-6,20H,(H,19,21)/b10-3-

InChI key

LMXYVLFTZRPNRV-KMKOMSMNSA-N

Gene Information

human ... RAF1(5894)

Application

GW5074 has been used as a specific inhibitor of cRaf1 in C2 murine skeletal myoblasts, HT-29 colorectal adenocarcinoma cell line and neutrophils.

Biochem/physiol Actions

GW5074 is a cRaf1 kinase inhibitor, which is placed immediately downstream of Ras in the Motogen activated protein kinase (MAPK) signaling pathway. It elicits protection in GW5074 prevents degeneration in Huntington′s disease and also have therapeutic potential in treating neurodegenerative disorders. GW5074 in combination with anti-inflammatory drug, dexamethasone, attenuates sidestream smoke-induced airway inflammation.

Other Notes

light sensitive

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The c-Raf inhibitor GW5074 provides neuroprotection in vitro and in an animal model of neurodegeneration through a MEK-ERK and Akt-independent mechanism
Chin PC, et al.
Journal of Neurochemistry, 90(3), 595-608 (2004)
Human intestinal epithelial cells respond to beta-glucans via Dectin-1 and Syk
Cohen-Kedar S, et al.
European Journal of Immunology, 44(12), 3729-3740 (2014)
Entamoeba histolytica induce signaling via Raf/MEK/ERK for neutrophil extracellular trap (NET) formation
Fonseca Z, et al.
Frontiers in Cellular and Infection Microbiology, 8(12), 226-226 (2018)
Yuan Zhang et al.
Scientific reports, 7(1), 10385-10385 (2017-09-06)
Enterovirus 71 (EV71) is a major pathogen of hand, foot, and mouth disease (HFMD). To date, no antiviral drug has been approved to treat EV71 infection. Due to the essential role that EV71 3 C protease (3C
Tomoko Nishimura et al.
PloS one, 3(7), e2558-e2558 (2008-07-04)
Selective serotonin reuptake inhibitors (SSRIs) have been widely used and are a major therapeutic advance in psychopharmacology. However, their pharmacology is quite heterogeneous. The SSRI fluvoxamine, with sigma-1 receptor agonism, is shown to potentiate nerve-growth factor (NGF)-induced neurite outgrowth in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service