Skip to Content
Merck
All Photos(4)

Key Documents

N0632

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide sodium salt

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N7NaO14P2
CAS Number:
Molecular Weight:
685.41
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥95% (HPLC)
≥95% (enzymatic)
≥95% (spectrophotometric assay)

form

powder

solubility

H2O: soluble-50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na].NC(=O)C1=CC=C[N](=C1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5c(N)ncnc45)C(O)C2O

InChI

1S/C21H28N7O14P2.Na.H/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;

InChI key

GSZRXOXBGUIRAU-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).
Electron acceptor

Other Notes

This is the common form of NAD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Villanueva et al.
Antioxidants & redox signaling, 30(18), 2013-2029 (2018-11-20)
Aims: The human apoptosis-inducing factor (hAIF) supports OXPHOS biogenesis and programmed cell death, with missense mutations producing neurodegenerative phenotypes.
Irshad Ali et al.
PloS one, 15(9), e0239340-e0239340 (2020-09-18)
In this study, an enzymatic pathway has been developed to replicate the Calvin Cycle by creating the individual steps of the carbon cycle in a bioreactor. The technology known as "artificial photosynthesis" converts CO2 emissions into a variety of intermediates
Lifeng Yang et al.
Cell metabolism, 31(4), 809-821 (2020-03-19)
NADH provides electrons for aerobic ATP production. In cells deprived of oxygen or with impaired electron transport chain activity, NADH accumulation can be toxic. To minimize such toxicity, elevated NADH inhibits the classical NADH-producing pathways: glucose, glutamine, and fat oxidation.
Mikako Yagi et al.
The EMBO journal, 40(8), e105268-e105268 (2021-02-03)
Mitochondrial translation dysfunction is associated with neurodegenerative and cardiovascular diseases. Cells eliminate defective mitochondria by the lysosomal machinery via autophagy. The relationship between mitochondrial translation and lysosomal function is unknown. In this study, mitochondrial translation-deficient hearts from p32-knockout mice were
Norbert Ha et al.
Nucleic acids research, 49(20), 11596-11613 (2021-11-02)
Using the programmable RNA-sequence binding domain of the Pumilio protein, we FLAG-tagged Xist (inactivated X chromosome specific transcript) in live mouse cells. Affinity pulldown coupled to mass spectrometry was employed to identify a list of 138 candidate Xist-binding proteins, from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service