Skip to Content
Merck
All Photos(3)

Documents

F9128

Sigma-Aldrich

5′-(4-Fluorosulfonylbenzoyl)adenosine hydrochloride

Synonym(s):

Adenosine-5′-(4-fluorosulfonylbenzoate) hydrochloride, FSBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16FN5O7S · HCl
CAS Number:
Molecular Weight:
489.86
Beilstein:
8178939
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥95% (TLC)

form

powder

solubility

methanol: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Cl[H].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COC(=O)c4ccc(cc4)S(F)(=O)=O)[C@@H](O)[C@H]3O

InChI

1S/C17H16FN5O7S.ClH/c18-31(27,28)9-3-1-8(2-4-9)17(26)29-5-10-12(24)13(25)16(30-10)23-7-22-11-14(19)20-6-21-15(11)23;/h1-4,6-7,10,12-13,16,24-25H,5H2,(H2,19,20,21);1H/t10-,12-,13-,16-;/m1./s1

InChI key

ZUHVYHQVJYIHPN-KHXPSBENSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5′-(4-Fluorosulfonylbenzoyl)adenosine (FSBA) is an ATP analog used to study the mechanism of action of wortmannin. FSBA has been used to study vesicular transport in intestinal cells such as Caco-2, and to covalently modify residues in the nucleotide-binding domains (NBDs) of various ATPases, kinases, and other proteins.
5′-(4-Fluorosulfonylbenzoyl)adenosine hydrochloride has been used to incubate with Ime2 (inducer of meiosis 2) to determine the sensitivity of Ime2 to the adenosine triphosphate (ATP) analog 5′-fluorosulfonylbenzoyladenosine (FSBA).

Biochem/physiol Actions

5′-(4-Fluorosulfonylbenzoyl)adenosine helps to recognize adenosine triphosphate (ATP)-binding sites in kinases due to its reaction with nucleophilic amino acids happening within these motifs.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Faudry et al.
The Journal of biological chemistry, 279(19), 19607-19613 (2004-02-27)
Apyrases are nucleoside triphosphate-diphosphohydrolases (EC 3.6.1.5) present in a variety of organisms. The apyrase activity found in the saliva of hematophagous insects is correlated with the prevention of ADP-induced platelet aggregation of the host during blood sucking. Purification of apyrase
Nicolas Menzel et al.
Analytical biochemistry, 368(2), 178-184 (2007-06-29)
Nearly all processes in cells are regulated by the coordinated interplay between reversible protein phosphorylation and dephosphorylation. Therefore, it is a great challenge to identify all phosphorylation substrates of a single protein kinase to understand its integration into intracellular signaling
Kenichi A Tanaka et al.
Platelets, 15(5), 325-332 (2004-09-17)
The use of clopidogrel (Plavix), an inhibitor of adenosine diphosphate (ADP)-induced platelet aggregation, has been proven to reduce ischemic events in cardiovascular patients, but little information is available for optimal monitoring of platelet function in patients receiving the drug preoperatively.
Steven J Ratcliffe et al.
Journal of biomolecular screening, 12(1), 126-132 (2006-12-15)
Most of the kinase inhibitors that are approved for therapeutic uses or that are undergoing clinical trials are directed toward the adenosine triphosphate (ATP) binding site of protein kinases. 5'-Fluorosulfonylbenzoyl 5'-adenosine (FSBA) is an activitybased probe (ABP) that covalently modifies
Giovanni Renzone et al.
Proteins, 67(1), 189-197 (2007-01-24)
Sso7d from the extreme thermophilic crenarchaeon Sulfolobus solfataricus is a multifunctional protein in in vitro assays, whose in vivo role is still puzzling. Crystals of Sso7d in complex with DNA elucidated the protein surface involved in the binding to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service