Skip to Content
Merck
All Photos(3)

Key Documents

62320

Sigma-Aldrich

(±)-α-Lipoic acid

≥98.0%

Synonym(s):

(±)-1,2-Dithiolane-3-pentanoic acid, 6,8-Dithiooctanoic acid, DL-α-Lipoic acid, DL-6,8-Thioctic acid, Lip(S2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
Beilstein:
81853
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Assay

≥98.0% (HPLC)
≥98.0%

form

solid

ign. residue

≤0.1%

loss

≤0.2% loss on drying

bp

>286 °C/1.013 hPa

mp

60-62 °C

solubility

water: 0.87 g/L at 22 °C

density

1.4 g/cm3 at 23 °C

functional group

carboxylic acid
disulfide

storage temp.

2-8°C

SMILES string

[OC(=O)CCCCC1CCSS1]
OC(=O)CCCCC1CCSS1

InChI

[1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)]
1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)

InChI key

[AGBQKNBQESQNJD-UHFFFAOYSA-N]
AGBQKNBQESQNJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

α-lipoic acid (ALA, thioctic acid) is a natural organosulfur compound produced in plants, animals and humans. It is a powerful antioxidant and plays a role in the forming nucleic acids, building blocks of DNA.

This product is a racemic mixture.

Application

(±)-α-Lipoic acid has been used:

  • In in vitro lipoylation studies and in the pyruvate dehydrogenase complex (PDC)-pyruvate dehydrogenase kinase (PDHK) functional assay.
  • To investigate its antioxidative effect on developing cerebellum of rats exposed to arsenic during postnatal period.

Biochem/physiol Actions

(±)-α-Lipoic acid exhibits direct free radical scavenging property and reduces the increased oxidative stress. In addition, it might also exhibit prooxidant activity. α-Lipoic acid functions as an important prosthetic group in α-keto acid dehydrogenase complexes of the mitochondria. It acts as a potential therapeutic for diabetic patients with distal symmetric polyneuropathy (DSP). α-Lipoic acid is also used in the treatment of energy-impaired and redox unbalanced diseases.
Antioxidant and coenzyme needed for the activity of enzyme complexes such as pyruvate dehydrogenase and glycine decarboxylase.
Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oral treatment with alpha-lipoic acid improves symptomatic diabetic polyneuropathy: the SYDNEY 2 trial
Ziegler D, et al.
Diabetes Care, 29(11), 2365-2370 (2006)
Catherine Grogan et al.
Sensors (Basel, Switzerland), 20(3) (2020-02-12)
The high sensitivity of silicon microcantilever sensors has expanded their use in areas ranging from gas sensing to bio-medical applications. Photochromic molecules also represent promising candidates for a large variety of sensing applications. In this work, the operating principles of
Antioxidant and prooxidant activities of alpha-lipoic acid and dihydrolipoic acid
Moini H, et al.
Toxicology and Applied Pharmacology, 182(1), 84-90 (2002)
Insights on the Use of ?-Lipoic Acid for Therapeutic Purposes
Salehi B., et. al.
Biomolecules, 9(8), 356-356 (2019)
Ameliorative role of antioxidant supplementation on sodium-arsenite induced adverse effects on the developing rat cerebellum
Kaushal P, et al.
Journal of Ayurveda and integrative medicine (2019)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service