Skip to Content
Merck
All Photos(1)

Key Documents

F8014

Sigma-Aldrich

Ferrichrome Iron-free from Ustilago sphaerogena

Synonym(s):

Deferriferrichrome

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45N9O12
CAS Number:
Molecular Weight:
687.70
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.25

storage temp.

2-8°C

Quality Level

SMILES string

C\C(CC(O)=O)=C/C1=[O][Fe]2345ON1CCCC6NC(=O)CNC(=O)C(CO)NC(=O)C(CO)NC(=O)C(CCCN(O2)C(\C=C(\C)CC(O)=O)=[O]3)NC(=O)C(CCCN(O4)C(\C=C(/C)CC(O)=O)=[O]5)NC6=O

InChI

1S/C27H45N9O12/c1-16(37)34(46)10-4-7-19-25(43)30-14-23(41)28-13-22(40)29-15-24(42)31-20(8-5-11-35(47)17(2)38)26(44)33-21(27(45)32-19)9-6-12-36(48)18(3)39/h19-21,46-48H,4-15H2,1-3H3,(H,28,41)(H,29,40)(H,30,43)(H,31,42)(H,32,45)(H,33,44)/t19-,20-,21-/m0/s1

InChI key

ZFDAUYPBCXMSBF-ACRUOGEOSA-N

Biochem/physiol Actions

Growth-promoting iron chelator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L Constantine et al.
Biopolymers, 30(3-4), 239-256 (1990-01-01)
We have applied computational procedures that utilize nmr data to model the solution conformation of ferrichrome, a rigid microbial iron transport cyclohexapeptide of known x-ray crystallographic structure [D. van der Helm et al. (1980) J. Am. Chem. Soc. 102, 4224-4231].
Sijing Wang et al.
Applied and environmental microbiology, 86(7) (2020-02-02)
The siderophore synthetic system in Shewanella species is able to synthesize dozens of macrocyclic siderophores in vitro with synthetic precursors. In vivo, however, although three siderophores are produced naturally in Shewanella algae B516, which carries a lysine decarboxylase (AvbA) specific
Microbial iron compounds.
J B Neilands
Annual review of biochemistry, 50, 715-731 (1981-01-01)
Takuya Iwama et al.
Cancer cell international, 21(1), 21-21 (2021-01-08)
Colorectal cancers develop through several pathways, including the adenoma-carcinoma sequence and colitis-associated carcinogenesis. An altered intestinal microflora has been reported to be associated with the development and progression of colorectal cancer via these pathways. We identified Lactobacillus casei-derived ferrichrome as
J D Herscheid et al.
Nuclear medicine communications, 7(1), 53-57 (1986-01-01)
The biodistribution of 67Ga-labelled complexes of some mono- and di-N-hydroxyamino acid derivatives and of two siderophores (trihydroxamates) were evaluated in Rhabdomyosarcoma-bearing rats. It was found that the mono- and dihydroxamates could not enhance the tumour-localizing properties of 67Ga-citrate, while the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service