Skip to Content
Merck
All Photos(1)

Key Documents

F5307

Sigma-Aldrich

5−Fluoro−2′−deoxycytidine

≥98% (HPLC), powder

Synonym(s):

2′-deoxy-5-fluoro-Cytidine, FCDR, FdCyd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN3O4
CAS Number:
Molecular Weight:
245.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

NC1=NC(=O)N(C=C1F)[C@H]2C[C@H](O)[C@@H](CO)O2

InChI

1S/C9H12FN3O4/c10-4-2-13(9(16)12-8(4)11)7-1-5(15)6(3-14)17-7/h2,5-7,14-15H,1,3H2,(H2,11,12,16)/t5-,6+,7+/m0/s1

InChI key

IDYKCXHJJGMAEV-RRKCRQDMSA-N

Biochem/physiol Actions

5-Fluoro-2′-deoxycytidine is a mechanism based DNMT (DNA cytosine-5 methyltransferase) inhibitor, that forms a covalent link with the cysteine residue in the active site of DNMT.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Shen et al.
Nucleic acids research, 23(21), 4275-4282 (1995-11-11)
DNA (cytosine-5)-methyltransferases can cause deamination of cytosine when the cofactor S-adenosylmethionine (AdoMet) is limiting and thus function as sequence-specific C-->U mutator enzymes. Here we explored whether mutations causing inactivation of the cofactor binding activity of the HpaII methyltransferase, thus mimicking
T Hanck et al.
Nucleic acids research, 21(2), 303-309 (1993-01-25)
The product of the dcm gene is the only DNA cytosine-C5 methyltransferase of Escherichia coli K-12; it catalyses transfer of a methyl group from S-adenosyl methionine (SAM) to the C-5 position of the inner cytosine residue of the cognate sequence
E Harrer et al.
The Journal of infectious diseases, 173(2), 476-479 (1996-02-01)
The human immunodeficiency virus (HIV) type 1 reverse transcriptase (RT) is an important target for therapeutic intervention and for HIV-1-specific cytotoxic T lymphocytes (CTL). An HLA-A2-restricted CTL epitope containing the sequence YMDD, which is highly conserved among human and animal
Theodore C Jessop et al.
Bioorganic & medicinal chemistry letters, 19(23), 6784-6787 (2009-10-20)
A series of deoxycytidine kinase inhibitors was simultaneously optimized for potency and PK properties. A co-crystal structure then allowed merging this series with a high throughput screening hit to afford a highly potent, selective and orally bioavailable inhibitor, compound 10.
M Iigo et al.
Cancer chemotherapy and pharmacology, 20(1), 1-4 (1987-01-01)
The plasma concentration of 5-fluorouracil (FUra) following the i.v. administration of FUra and guanosine 5'-monophosphate (GMP) or guanosine 5'-triphosphate (GTP) was markedly elevated. These values were more than 5-fold higher than those obtained with FUra alone over 60 min after

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service