Skip to Content
Merck
All Photos(10)

Key Documents

B5002

Sigma-Aldrich

5-Bromo-2′-deoxyuridine

≥99% (HPLC)

Synonym(s):

5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O5
CAS Number:
Molecular Weight:
307.10
Beilstein:
30395
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥99% (HPLC)

form

powder

mp

191-194 °C (dec.) (lit.)

solubility

DMSO: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(Br)C(=O)NC2=O

InChI

1S/C9H11BrN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

WOVKYSAHUYNSMH-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-2′-deoxyuridine (5-BrdU) is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.
5-Bromo-2′-deoxyuridine has been used:
  • as a supplement to study the effect of electroconvulsive seizures on hippocampal neurogenesis
  • as a supplement to study the loss-of-gene function in adult zebrafish heart
  • in T lymphocyte proliferation assay{133]

Biochem/physiol Actions

5-Bromo-2′-deoxyuridine (BrdU) is used to analyze cell proliferation, because of its facile incorporation into DNA during the S phase of the cell cycle. BrdU stimulates cellular differentiation and maturation in leukemia cell lines, while it inhibits differentiation of friend erythroleukemia cells. Studies of BrdU incorporation often subsequently use BrdU-specific antibodies with fluorescent tags, for detection of the incorporated BrdU, via such methods as flow cytometry or fluorescence microscopy.
Thymidine analog used as a mutagen in genetic research. Selectively incorporated into cellular DNA during S-phase.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PEG-PLA nanoparticles facilitate siRNA knockdown in adult zebrafish heart
Diao J, et al.
Developmental Biology, 406, 196-202 (2015)
Mechanisms of Differentiation, Volume 2, 85-85 (1990)
Target Validation in Drug Discovery, 109-109 (2011)
Electroconvulsive seizures increase hippocampal neurogenesis after chronic corticosterone treatment
Hellsten J, et al.
The European Journal of Neuroscience, 16(2), 283-290 (2002)
Francisco Alén et al.
International journal of molecular sciences, 20(14) (2019-07-25)
Mutant mice with respect to the splicing factor Zrsr1 present altered spermatogenesis and infertility. To investigate whether Zrsr1 is involved in the homeostatic control that the hypothalamus exerts over reproductive functions, we first analyzed both differential gene and isoform expression

Articles

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service