Skip to Content
Merck
All Photos(1)

Documents

D9026

Supelco

Digoxigenin

analytical standard

Synonym(s):

3β,12β,14β,21-Tetrahydroxy-20(22)-norcholenic acid lactone, 3β,12β,14-Trihydroxy-5β,20(22)-cardenolide, 5β,20(22)-Cardenolide-3β,12β,14-triol, Lanadigigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
Beilstein:
96479
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

Assay

≥98% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

222 °C (lit.)

application(s)

environmental
food and beverages
forensics and toxicology
veterinary

format

neat

functional group

ketone

storage temp.

room temp

SMILES string

C[C@]12CC[C@H](O)C[C@H]1CC[C@@H]3[C@@H]2C[C@@H](O)[C@]4(C)[C@H](CC[C@]34O)C5=CC(=O)OC5

InChI

1S/C23H34O5/c1-21-7-5-15(24)10-14(21)3-4-17-18(21)11-19(25)22(2)16(6-8-23(17,22)27)13-9-20(26)28-12-13/h9,14-19,24-25,27H,3-8,10-12H2,1-2H3/t14-,15+,16-,17-,18+,19-,21+,22+,23+/m1/s1

InChI key

SHIBSTMRCDJXLN-KCZCNTNESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Digoxigenin is a therapeutic drug belonging to the group of cardiac glycosides, widely used in the management of congestive heart failure and other cardiac diseases.

Application

Digoxigenin is used as an analytical reference standard for the quantification of the analyte in biological samples using high-performance liquid chromatography couped to tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Acute Tox. 2 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and Quantification of Cardiac Glycosides in Blood and Urine Samples by HPLC/MS/MS
Guan F, et al.
American Chemical Science Journal (1999)
Simone Dealtry et al.
Applied and environmental microbiology, 80(13), 4012-4020 (2014-04-29)
Biopurification systems (BPS) are used on farms to control pollution by treating pesticide-contaminated water. It is assumed that mobile genetic elements (MGEs) carrying genes coding for enzymes involved in degradation might contribute to the degradation of pesticides. Therefore, the composition
Vicki Adams et al.
Applied and environmental microbiology, 80(12), 3597-3603 (2014-04-01)
TnpX is a site-specific recombinase responsible for the excision and insertion of the transposons Tn4451 and Tn4453 in Clostridium perfringens and Clostridium difficile, respectively. Here, we exploit phenotypic features of TnpX to facilitate genetic mutagenesis and complementation studies. Genetic manipulation
Christine E Tinberg et al.
Nature, 501(7466), 212-216 (2013-09-06)
The ability to design proteins with high affinity and selectivity for any given small molecule is a rigorous test of our understanding of the physiochemical principles that govern molecular recognition. Attempts to rationally design ligand-binding proteins have met with little
Suzanne E McGaugh et al.
Nature communications, 5, 5307-5307 (2014-10-21)
Natural populations subjected to strong environmental selection pressures offer a window into the genetic underpinnings of evolutionary change. Cavefish populations, Astyanax mexicanus (Teleostei: Characiphysi), exhibit repeated, independent evolution for a variety of traits including eye degeneration, pigment loss, increased size

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service