Skip to Content
Merck
All Photos(2)

Key Documents

F0896

Sigma-Aldrich

5-Fluoro-DL-tryptophan

≥95% (HPLC)

Synonym(s):

5-fluoro-tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11FN2O2
CAS Number:
Molecular Weight:
222.22
Beilstein:
22753
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

5-Fluoro-DL-tryptophan, powder or crystals

Assay

≥95% (HPLC)

Quality Level

form

powder or crystals

color

slightly off-white to brown

application(s)

detection
peptide synthesis

storage temp.

2-8°C

SMILES string

NC(Cc1c[nH]c2ccc(F)cc12)C(O)=O

InChI

1S/C11H11FN2O2/c12-7-1-2-10-8(4-7)6(5-14-10)3-9(13)11(15)16/h1-2,4-5,9,14H,3,13H2,(H,15,16)

InChI key

INPQIVHQSQUEAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Determination of the 19F NMR chemical shielding tensor and crystal structure of 5-fluoro-dl-tryptophan.: This study provides a detailed analysis of the 19F NMR chemical shielding tensor and the crystal structure of 5-Fluoro-dl-tryptophan, offering valuable insights into its electronic environment and molecular interactions. Such detailed structural characterization is crucial for the development of fluorinated drugs and biomolecules, enhancing our understanding of their interactions and functionalities (Sykes BD et al., 2007).

Biochem/physiol Actions

5-Fluoro-DL-tryptophan (5-F-TRP), a fluorine-containing analogue of tryptophan, may be incorporated into proteins such as cyclic AMP receptor protein (CRP) from Escherichia coli to study changes in ligand binding. 5-F-TRP may be used as an internal reference for chromatographic analysis of tryptophan and its derivatives found in whole blood.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kinoshita et al.
Journal of bioscience and bioengineering, 92(6), 556-559 (2005-10-20)
In our study on nutritional requirement for the hyphal growth of Schizophyllum commune, we found that a Trp- mutant could not grow in the L-Trp-supplied medium in the presence of L-Ser. Further growth studies showed that not only L-Ser but
F Sixl et al.
The Biochemical journal, 266(2), 545-552 (1990-03-01)
Two fluorine-containing analogues of the cyclic AMP receptor protein (CRP) from Escherichia coli were prepared by biosynthetic incorporation of 5-fluorotryptophan (5-F-Trp) and 3-fluorotyrosine (3-F-Tyr). The 19F-n.m.r. spectrum of the [5-F-Trp]CRP showed two signals corresponding to the two tryptophan residues, and
N Aymard et al.
Progress in neuro-psychopharmacology & biological psychiatry, 18(1), 77-86 (1994-01-01)
1. 5-Hydroxytryptamine (5-HT) is a well known neurotransmitter implicated in mental disorders, tryptophan (Trp) as amino acid precursor may be interesting. 2. A rapid and simple reversed-phase (mu Bondapac C18 as stationary phase) liquid chromatographic method with fluorimetric detection (excitation:
Kenji Hamase et al.
Journal of chromatography. A, 1106(1-2), 159-164 (2006-01-31)
A sensitive assay for D-amino-acid oxidase (DAO) activity in mammalian tissues has been established. D-Tryptophan (D-Trp) analogs were tested as substrates for DAO, and 5-fluoro-D-tryptophan (D-FTP) was found to be the best substrate. By the enzymatic reaction, D-FTP was converted
S Rozovsky et al.
Journal of molecular biology, 310(1), 271-280 (2001-06-23)
Product release is partially rate determining in the isomerization reaction catalyzed by Triosephosphate Isomerase, the conversion of dihydroxyacetone phosphate to D-glyceraldehyde 3-phosphate, probably because an active-site loop movement is necessary to free the product from confinement in the active-site. The

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service