Skip to Content
Merck
All Photos(1)

Key Documents

T2610000

Tryptophan

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Tryptophan, (S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86197
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

tryptophan

manufacturer/tradename

EDQM

mp

280-285 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1

InChI key

QIVBCDIJIAJPQS-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tryptophan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Amino acid precursor of serotonin and melatonin
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Platten et al.
Cancer research, 72(21), 5435-5440 (2012-10-24)
Tryptophan catabolism in cancer is increasingly being recognized as an important microenvironmental factor that suppresses antitumor immune responses. It has been proposed that the essential amino acid tryptophan is catabolized in the tumor tissue by the rate-limiting enzyme indoleamine-2,3-dioxygenase (IDO)
Annemieke T van der Goot et al.
Trends in molecular medicine, 19(6), 336-344 (2013-04-09)
Aging is an important risk factor for many debilitating diseases, including cancer and neurodegeneration. In model organisms, interfering with metabolic signaling pathways, including the insulin/insulin-like growth factor (IGF) 1 (IIS) and TOR pathways, can protect against age-related pathologies and increase
M Diksic
Journal of psychiatry & neuroscience : JPN, 26(4), 293-303 (2001-10-10)
The alpha-methyl-L-trypotophan (alpha-MTrp) method for the study of the brain serotonergic system is based on the fact that labelled alpha-MTrp is taken up by and, in part, retained in the brain, and this retention (trapping) is proportional to brain serotonin
M Diksic et al.
Journal of neurochemistry, 78(6), 1185-1200 (2001-10-02)
alpha-Methyl-L-tryptophan (alpha-MTrp) is an artificial amino acid and an analog of tryptophan (Trp), the precursor of the neurotransmitter serotonin (5-HT). In this article we have summarized available data, which suggest that the measurement of the unidirectional uptake of alpha-MTrp and
Simon N Young
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1615), 20110375-20110375 (2013-02-27)
Acute tryptophan depletion (ATD) studies indicate that low serotonin can lower mood and also increase aggression, although results vary somewhat between studies with similar participants. Lowering of mood after ATD is related to the susceptibility of the study participants to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service