Skip to Content
Merck
All Photos(1)

Documents

93659

Sigma-Aldrich

L-Tryptophan

≥99.5% (NT), BioUltra

Synonym(s):

(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86197
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Tryptophan, BioUltra, ≥99.5% (NT)

product line

BioUltra

Quality Level

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D −31.5±1°, c = 1% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.1%

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

mp

280-285 °C (dec.) (lit.)

solubility

0.5 M HCl: 0.1 g/mL at 20 °C, clear, colorless to faintly yellow

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 326 nm in 0.5 M HCl at 0.5 M

application(s)

cell analysis

SMILES string

N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1

InChI key

QIVBCDIJIAJPQS-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Tryptophan has been used as a nutritional supplement to study its effect on the circadian clock of primary cell lines. It has also been used as a test amino acid aerosol to study the variation in the fluorescence intensity of thermally-exposed bacterial bioaerosols.

Biochem/physiol Actions

Tryptophan (Trp) is one of the functional amino acids that are associated with growth, reproduction, maintenance and immunity. Increased Trp availability is necessary for the regulation of mood, cognition and behaviour. It is hypothesised that L-Trp might be useful in inducing sleep in healthy adults against the normal circadian rhythm. Trp uptake by the brain depends on the plasma ratio of Trp to all of the other LNAAs (large neutral amino acids). Higher the Trp:LNAAs ratio, greater is the Trp uptake.

Other Notes

Amino acid precursor of serotonin and melatonin
In tryptophan-substituted Sephadex® and Sepharose for the improved chromatographic separation of cellulases; As spacer in isotachophoresis of serum proteins; Growth requirement of various microorganisms.

Legal Information

Sephadex is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Microbiology, 4, 1-1 (1974)
Long-lasting effect of perinatal exposure to L-tryptophan on circadian clock of primary cell lines established from male offspring born from mothers fed on dietary protein restriction.
Nascimento E
PLoS ONE, 8(2), 1-14 (2013)
T. Hine et al.
Electrophoresis ?83, 541-541 (1984)
M A Vijayalakshmi et al.
Journal of chromatography, 177(2), 201-208 (1979-09-21)
The adsorption of tyrosine, tryptophan and some related compounds on tryptophan and tryptophanyltryptophan gels is similar to but much stronger than that observed with the parent gel Sephadex G-25. The adsorption has been attributed mainly to charge transfer or redistribution
Variation in the fluorescence intensity of thermally-exposed bacterial bioaerosols
Jae HeeJung and Jung EunLee
Journal of Aerosol Science, 65, 101-110 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service