Skip to Content
Merck
All Photos(2)

Key Documents

D4139

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine dihydrochloride

≥99.0% purity (titration), powder

Synonym(s):

4-Amino-N,N-dimethylaniline dihydrochloride, DMPD · 2HCl, DMPPDA · 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2 · 2HCl
CAS Number:
Molecular Weight:
209.12
Beilstein:
3913463
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

N,N-Dimethyl-p-phenylenediamine dihydrochloride, suitable for peroxidase test, ≥99.0% (titration)

Quality Level

Assay

≥99.0% (titration)

form

powder

technique(s)

titration: suitable

color

white to light pink, and tan

mp

222 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear

suitability

suitable for peroxidase test

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].Cl[H].CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.2ClH/c1-10(2)8-5-3-7(9)4-6-8;;/h3-6H,9H2,1-2H3;2*1H

InChI key

IAEDWDXMFDKWFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethyl-p-phenylenediamine dihydrochloride (DMPD) has been used in the HID-AB (high iron diamine-alcian blue) staining of intestinal tract samples.

Biochem/physiol Actions

N,N-Dimethyl-p-phenylenediamine dihydrochloride (DMPD) is used for the determination of peroxidase. DMPD is converted into a red pigment of semiquinone nature. It is also used in the HID-AB (high iron diamine-alcian blue) staining procedures, which are used to differentiate between sialomucin and sulfomucin in the gastrointestinal tract.

Caution

Darkens readily when exposed to air.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for serial tissue processing and parallel analysis of aberrant crypt morphology, mucin depletion, and Beta-catenin staining in an experimental model of colon carcinogenesis.
McGinley JN, et al.
Biological Procedures Online, 12, 9032-9032 (2010)
Digestive Disease Pathology (2013)
Colorimetric analysis with N, N-dimethyl-p-phenylenediamine of the uptake of intravenously injected horseradish peroxidase by various tissues of the rat.
STRAUS W
The Journal of Biophysical and Biochemical Cytology, 4, 541-541 (1958)
The distribution of mucous secreting cells in the gastrointestinal tracts of three small rodents from Saudi Arabia: Acomys dimidiatus, Meriones rex and Meriones libycus.
Johnson O, et al.
Acta Histochemica, 118, 118-118 (2016)
Claudineia R Silva et al.
Talanta, 85(3), 1703-1705 (2011-08-03)
A spectrophotometric flow injection procedure involving N,N-dimethyl-p-phenylenediamine (DMPD) is applied to the sulfide monitoring of a sugar fermentation by Saccharomyces cerevisiae under laboratory conditions. The gaseous chemical species evolving from the fermentative process, mainly CO(2), are trapped allowing a cleaned

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service