Skip to Content
Merck
All Photos(1)

Key Documents

07770

Millipore

N,N-Dimethyl-p-phenylenediamine dihydrochloride

suitable for microbiology, ≥99.0%

Synonym(s):

4-Amino-N,N-dimethylaniline dihydrochloride, DMPD · 2HCl, DMPPDA · 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2 · 2HCl
CAS Number:
Molecular Weight:
209.12
Beilstein:
3913463
EC Number:
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (AT)
≥99.0%

form

powder

loss

≤0.05% loss on ignition

mp

210-215 °C (dec.)
222 °C (dec.) (lit.)

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

food and beverages
microbiology

SMILES string

Cl[H].Cl[H].CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.2ClH/c1-10(2)8-5-3-7(9)4-6-8;;/h3-6H,9H2,1-2H3;2*1H

InChI key

IAEDWDXMFDKWFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyl-p-phenylenediamine dihydrochloride is an azoreduction product of acid orange 52 (AO52). It is useful in the oxidase test which differentiates bacteria based on their ability to utilise the dye.

Application

N,N-Dimethyl-p-phenylenediamine dihydrochloride has been used to determine the plasma oxidant capacity in terms of plasma ferric equivalent oxidative potential (PFEOP).

Caution

May discolor to red on storage.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Alexander
Encyclopedia of Microbiology (2000)
Modulation of Erythrocyte Plasma Membrane Redox System Activity by Curcumin
Prabhakar Singh
Biochemistry Research International (2016)
Veronica Verde et al.
Free radical research, 36(8), 869-873 (2002-11-08)
Oxidative stress has been clearly implicated in human disease by a growing body of scientific evidences. There is no ideal method for the measurement of this parameter. A possible strategy would be to measure simultaneously several biomarkers representing damage to
F Tessier et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 66(2), 166-170 (1996-01-01)
An HPLC micro-method with fluorescence detection has been developed to determine total vitamin C (vit C) and dehydroascorbic acid (DHA) concentrations in human plasma samples. This method is based on the rapid, specific reaction of DHA with dimethyl-o-phenylenediamine (DMPD) to
S Marroquí et al.
Journal of bacteriology, 183(3), 854-864 (2001-02-24)
We isolated a Tn5-induced Rhizobium tropici mutant that has enhanced capacity to oxidize N,N-dimethyl-p-phenylendiamine (DMPD) and therefore has enhanced respiration via cytochrome oxidase. The mutant had increased levels of the cytochromes c(1) and CycM and a small increase in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service