Skip to Content
Merck
All Photos(1)

Key Documents

193992

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine

97%

Synonym(s):

4-(Dimethylamino)aniline, 4-Amino-N,N-dimethylaniline, N,N-Dimethyl-1,4-phenylenediamine, DMPPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2
CAS Number:
Molecular Weight:
136.19
Beilstein:
508105
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Assay

97%

form

solid

bp

262 °C (lit.)

mp

34-36 °C (lit.)

SMILES string

CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2/c1-10(2)8-5-3-7(9)4-6-8/h3-6H,9H2,1-2H3

InChI key

BZORFPDSXLZWJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyl-p-phenylenediamine (DPD, DMPD) is an aromatic amine mainly used as an intermediate to produce dyes. Its oxidation reaction with H2O2 in the presence of iron(III) catalyst has been used for the spectrophotometric detection of trace quantities of iron (III). Cyclovoltammetric studies have been conducted to evaluate the electrochemical oxidation of DMPD at various pH. DMPD on oxidation under basic conditions has been reported to undergo hydrolytic decomposition.

Application

N,N-Dimethyl-p-phenylenediamine may be used for the preparation of molecular compounds with tetracyano-p-quinodimethane (TCNQ). Polarized absorption spectra of these molecular compounds were investigated.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric catalytic determination of an ultratrace amount of iron (III) in water based on the oxidation of N, N-dimethyl-p-phenylenediamine by hydrogen peroxide.
Hirayama K and Unohara N.
Analytical Chemistry, 60(23), 2573-2577 (1988)
Polarized Absorption Spectra of Single Crystals of Ion Radical Salts. I. Molecular Compounds of 7, 7, 8, 8-Tetracyano-p-quinodimethane with N, N, N', N'-Tetramethyl-p-phenylenediamine and N, N-Dimethyl-p-phenylenediamine.
Kuroda H, et al.
Bulletin of the Chemical Society of Japan, 41(12), 2855-2858 (1968)
Mechanism diversity in anodic oxidation of N, N-dimethyl-p-phenylenediamine by varying pH.
Maleki A and Nematollahi D.
Journal of Electroanalytical Chemistry, 704, 75-79 (2013)
An historical review of the use of dye precursors in the formulation of commercial oxidation hair dyes.
Corbett JF.
Dyes and Pigments, 41(1), 127-136 (1999)
C Li et al.
The Biochemical journal, 300 ( Pt 1), 31-36 (1994-05-15)
C-terminal alpha-amidation is a structural feature essential to the biological activity of many peptide hormones. Peptidylglycine alpha-amidating mono-oxygenase (PAM; EC 1.14.17.3) catalyses conversion of glycine-extended peptide hormone precursors into their corresponding alpha-hydroxyglycine derivatives. This reaction is the first step in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service