Skip to Content
Merck
All Photos(2)

Documents

C7821

Sigma-Aldrich

Curdlan

from Alcaligenes faecalis, ≥98% (TLC), solid

Synonym(s):

β-1,3-Glucan hydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Curdlan from Alcaligenes faecalis,

form

solid

Quality Level

storage temp.

2-8°C

General description

Curdlan is an insoluble natural polymer with linear structure mainly produced by Alcaligenes faecalis. It is a dietary fiber with gelatin properties.

Application

Curdlan from Alcaligenes faecalis has been used:
  • for the stimulation of carp macrophages for regulation of differentially expressed genes
  • to stimulate human intestinal epithelial cells for cytokine secretion
  • to study its effect on transcriptional responses of inflammation/respiratory burst-associated genes in mouse alveolar macrophages by customized reverse transcription (RT)-PCR based arrays
  • as a substrate in β-1,3–1,4-Glucanase activity assay

Biochem/physiol Actions

Curdlan is used as a food additive in the food industry. It improves the elasticity and reduces the embrittling tendency of hydroxyapatite/chitosan scaffold, which aids in bone regeneration.
Curdlan is a (1,3)-β-glucan. It is a pathogen-associated molecular-pattern (PAMP) recognized by the receptor dectin-1. Activation of dectin-1, together with TLR2/TLR6, promotes signaling resulting in the production of pro-inflammatory cytokines, thus inducing inflammation.

Preparation Note

For gelation at 70 °C, the concentration of the sodium hydroxide should be 0.01 M.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of natural polymer-based hydrogels in the food industry
"Handbook of Poylmer-Liquid Interaction Parameters and Solubility Parameters ", 357-410 (2020)
"Reference Module in Food Science (2020)
K N Masihi et al.
International journal of immunopharmacology, 19(9-10), 463-468 (1998-06-24)
The effects of glucan-based immunomodulators curdlan sulfate (CRDS) and lentinan on cytokine production stimulated by lipopolysaccharide (LPS) in bacillus Calmette-Guerin (BCG)-primed mice were investigated. Pretreatment with CRDS or lentinan before LPS administration induced a striking inhibition of up to 89%
Jules Petit et al.
Frontiers in immunology, 10, 280-280 (2019-03-14)
Immune-modulatory effects of β-glucans are generally considered beneficial to fish health. Despite the frequent application of β-glucans in aquaculture practice, the exact receptors and downstream signalling remains to be described for fish. In mammals, Dectin-1 is a member of the
Nicola Jackson et al.
European journal of immunology, 44(4), 962-969 (2014-01-25)
Type B T cells recognize peptide-MHC class II (pMHCII) isoforms that are structurally distinct from those recognized by conventional type A T cells. These alternative type B conformers result from peptide loading in the absence of HLA-DM. Type A conformers

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service