Skip to Content
Merck

Skip To

C9752

Chitin from shrimp shells

BioReagent, suitable for analysis of chitinase, purified powder

Synonym(s):

Poly-(1→4)-β-N-acetyl-D-glucosamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
215-744-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

shrimp (Pandalus borealis)

Quality Segment

product line

BioReagent

form

purified powder

color

yellow to tan

suitability

suitable for analysis of chitinase

SMILES string

CC(=O)N[C@@H]1[C@H](O)O[C@@H](CO)[C@H](O)[C@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4?,5?,6?,7?,8-/m1/s1

InChI key

OVRNDRQMDRJTHS-WTZNIHQSSA-N

Application

Chitin from shrimp shells, a long-chain polymer of a β-1,4 linked N-acetylglucosamine residues, may be used to study its physicochemical properties and role in the formation of arthropod exoskeletons. It may be used to identify and characterize chitinase(s) and chitin deacetylase(s), as a standard to study the structure of chitins from other species and as a viscosity/thickening agent or binder. Chitin from shrimp shells may be used in chitin asthma and allergy research.

Preparation Note

Purified by a modification of the method of Hirano, S. and Nagao, N., Agric. Biol. Chem., 52, 2111 (1988).

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7170417963C3646
description

BioReagent, suitable for analysis of chitinase, purified powder

description

practical grade, powder

description

from shrimp shells, practical grade

description

from shrimp shells, ≥75% (deacetylated)

biological source

shrimp (Pandalus borealis)

biological source

shrimp (Pandalus borealis)

biological source

shrimp shells

biological source

shrimp shells

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

purified powder

form

powder

form

solid

form

powder or flakes

suitability

suitable for analysis of chitinase

suitability

-

suitability

-

suitability

-

product line

BioReagent

product line

-

product line

-

product line

-

color

yellow to tan

color

white to beige

color

-

color

white to beige


Still not finding the right product?

Explore all of our products under Chitin from shrimp shells


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. How can I dissolve chitin for long-term storage? Which solvent should I use?

    1 answer
    1. The solution stability of this product has not been determined. Some sources indicate that storage at -80°C for 1 year is possible. This product is incompatible with strong oxidizing agents. It can be dissolved in 0.1 M sodium acetate at pH 6.0 with a concentration of 5 mg/ml, resulting in a turbid faint yellow/tan solution that may contain insolubles.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. What is molecular weight of this product?

    1 answer
    1. Unfortunately, the molecular weight is not determined for this product.

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters