Skip to Content
Merck
All Photos(1)

Key Documents

A3007

Sigma-Aldrich

N-Acetylmuramic acid

≥98% (TLC)

Synonym(s):

(R)-2-(Acetylamino)-3-O-(1-carboxyethyl)-2-deoxy-D-glucose, 2-Acetamido-2-deoxy-3-O-(D-1-carboxyethyl)-D-glucopyranose, MurNAc, NAMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO8
CAS Number:
Molecular Weight:
293.27
Beilstein:
1994245
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤1 mol/mol methanol

color

white

mp

125  °C ((257 °F ))

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CC(O[C@H]1[C@H](O)[C@@H](CO)OC(O)[C@@H]1NC(C)=O)C(O)=O

InChI

1S/C11H19NO8/c1-5(11(18)19)20-10(9(17)8(16)4-14)7(3-13)12-6(2)15/h3,5,7-10,14,16-17H,4H2,1-2H3,(H,12,15)(H,18,19)/t5-,7+,8-,9-,10-/m1/s1

InChI key

SOARVSUSWULNDI-TVVSKHENSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetylmuramic acid (NAMA), a lactic acid ether derivative of N-acetylglucosamine found in bacterial cell wall proteoglycans, is used as a substrate to identify, differentiate and characterize N-acetylmuramic acid/N-acetylglucosamine kinase(s) and N-acetylmuramic acid etherase(s).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weidong Zhu et al.
Journal of periodontal & implant science, 46(1), 2-9 (2016-03-05)
Porphyromonas gingivalis and Tannerella forsythia have been implicated as the major etiologic agents of periodontal disease. These two bacteria are frequently isolated together from the periodontal lesion, and it has been suggested that their interaction may increase each one's virulence
Emma Hernandez-Sanabria et al.
Frontiers in cellular and infection microbiology, 7, 235-235 (2017-06-24)
Understanding the driving forces behind the shifts in the ecological balance of the oral microbiota will become essential for the future management and treatment of periodontitis. As the use of competitive approaches for modulating bacterial outgrowth is unexplored in the
Tingjun Liu et al.
Frontiers in cellular and infection microbiology, 10, 344-344 (2020-08-01)
Streptococcus mutans is an oral species closely associated with dental caries. As an early oral colonizer, S. mutans utilizes interspecies coaggregation to promote the colonization of subsequent species and affect polymicrobial pathogenesis. Previous studies have confirmed several adhering partner species
C S Hughes et al.
Biochimica et biophysica acta, 1861(8), 1951-1959 (2017-05-18)
A-type resistance towards "last-line" glycopeptide antibiotic vancomycin in the leading hospital acquired infectious agent, the enterococci, is the most common in the UK. Resistance is regulated by the VanR
Justin A Hutcherson et al.
Scientific reports, 7(1), 10474-10474 (2017-09-07)
The oral microflora is composed of both health-promoting as well as disease-initiating bacteria. Many of the disease-initiating bacteria are anaerobic and include organisms such as Porphyromonas gingivalis, Prevotella intermedia, Fusobacterium nucleatum, and Tannerella forsythia. Here we investigated a novel therapeutic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service