Skip to Content
Merck
All Photos(3)

Key Documents

C7762

Sigma-Aldrich

α-Chymotrypsin from bovine pancreas

Type I-S, essentially salt-free, lyophilized powder

Synonym(s):

α-chymotrypsin A and B, alpha-chymotrypsin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010112
NACRES:
NA.54

type

Type I-S

Quality Level

form

essentially salt-free, lyophilized powder

specific activity

≥40 units/mg protein

mol wt

25 kDa

purified by

3× crystallization

solubility

1 mM HCl: soluble 2.0 mg/mL, clear

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

α-Chymotrypsin from bovine pancreas has been used in a study to investigate protein extraction by Winsor-III microemulsion systems. α-Chymotrypsin from bovine pancreas has also been used in a study to investigate a new specific fullerene-based fluorescent probe for trypsin.
The product has been used to investigate the inhibitory effect of several ether oligomers against the enzyme. It has also been used to cleave pro-phenoloxidase in order to estimate total phenoloxidase in haemolymph. Furthermore, the enzyme has been used as a positive control in chymotrypsin assays using salivary gland and anterior midgut extracts of Deraeocoris nigritulus.

Biochem/physiol Actions

α-Chymotrypsin is a serine peptidase and has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. Molecular weight of this enzyme is found to be 25 kDa. The pI is 8.75. It selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan, and leucine. Ca2+ activates and stabilizes the enzyme. The enzyme is inhibited by diisopropyl fluorophosphate (DFP), phenylmethanesulfonyl fluoride (PMSF), N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK), chymostatin, aprotinin, α1-antitrypsin, and α2-macroglobulin, as well as 10 mM of Cu2+ and Hg2+.
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met, Leu) on the carboxyl end of the bond.

Unit Definition

One unit will hydrolyze 1.0 μmole of BTEE per min at pH 7.8 at 25 °C.

Preparation Note

Prepared free of autolysis products and low molecular weight contaminants.
The powder may be reconstituted in 1 mM HCl at 2 mg/mL concentration to form a clear solution.

Analysis Note

Minimum 85% protein
Protein determined by E1%/280

Other Notes

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Renaudet et al.
Organic letters, 6(3), 397-400 (2004-01-30)
[structure: see text] Hydroxyaromatic aldehydes and ketones were used as building blocks to prepare ether oligomers. An iterative two-step protocol involving Mitsunobu coupling and carbonyl reduction provided a protecting-group-free route with high yields. Activity screening of an 84-member library against
Nathan W Bailey et al.
Biology letters, 7(2), 217-220 (2010-09-03)
Increased investment in immunity is expected to be beneficial under crowded conditions because of the greater risk of pathogen and parasite transmission, but the evolution of this facultative response relies on the ability to accurately assess social cues in the
J A Hodgson et al.
Journal of general microbiology, 131(12), 3219-3227 (1985-12-01)
The effects of elevated temperature and of digestion with a variety of proteinases on the flocforming ability of flocculent strains of Saccharomyces cerevisiae, both genetically defined (FLO1 and FLO5) laboratory and genetically undefined brewing strains, have been determined. This has
Lakshmi Swarnalatha Jasti et al.
Biotechnology progress, 30(2), 317-323 (2014-01-23)
Allyl glycidyl ether (AGE)-ethylene glycol dimethacrylate (EGDM) copolymer with 25% crosslink density (AGE-25) shows excellent bovine serum albumin (BSA) adsorption (up to 16% (w/w)) at pH 8.0 and the adsorbed BSA is strongly bound. This protein-coated polymer provides a novel
Digestive enzymes and stylet morphology of Deraeocoris nigritulus (Uhler)(Hemiptera: Miridae) reflect adaptations for predatory habits.
Boyd Jr, D. W.
Annals of the Entomological Society of America, 96(5), 667-671 (2003)

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Protocols

Follow our procedure for the determination of a chymotrypsin activity. This enzymatic assay of alpha chymotrypsin guides you through the entire process and necessary calculations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service